Alginic acid, sodium salt - FAQs
View additional product information for Alginic acid, sodium salt - FAQs (177770050, 177775000, 177770025, 177772500)
1 product FAQs found
What is the molecular weight and M/G ratio of Alginic acid, sodium salt (Cat. No. 177775000, 177770050, 177772500, 177770025)?
The molecular weight of Alginic acid, sodium salt (Cat. No. 177775000, 177770050, 177772500, 177770025) is 450,000-550,000 Dalton and the M/G ratio is 1.5-1.6, thus ~60% mannuronic acid (m) and ~40% guluronic acid (n).