Search
View additional product information for Sodium butyrate, 98+% - FAQs (A11079.36, A11079.06, A11079.22)
1 product FAQs found
Sodium butyrate is soluble in water (0.1 g/mL), DMSO (<1 mg/mL), ethanol (~5 mg/mL), and PBS pH7. 2 (~10 mg/mL). Sodium butyrate is insoluble in ether.