Search
View additional product information for CyQUANT™ LDH and G6PD Cytotoxicity Assays - FAQs (V23111, C20301, C20300, C20303, C20302)
4 product FAQs found
The CyQUANT LDH Cytotoxicity Assay - Fluorescence Kit (Cat. No. C20302) contains sufficient reagents to perform 200 assays in 96-well plates.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Here are potential causes:
- Fluorescence may be quenched and/or LDH activity may be inhibited by various chemical compounds.
- The LDH may be precipitated/denatured.
- LDH can also be digested by any proteases liberated in the extracellular medium, examine the samples for protease activity.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Yes. The Spontaneous LDH Release Control provides the lower limit of natural LDH release for untreated samples (healthy cells). Without this control, you cannot determine whether any treatment resulted in any change in release, which relates to levels of cytotoxicity.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
We do not recommend doing this. LDH is an enzyme that will lose activity in diluted form when stored over time, refrigeration would not limit this loss of activity and freezing would cause a greater loss of activity.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.