Technical Resources - Media Formulations
13055 - TC-100
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.0650.08.666667
L-Alanine89.0225.02.52809
L-Arginine174.0550.03.1609194
L-Asparagine132.0350.02.6515152
L-Aspartic acid133.0350.02.631579
L-Cysteine121.020.00.16528925
L-Glutamic Acid147.0600.04.0816326
L-Glutamine146.0600.04.109589
L-Histidine155.03400.021.935484
L-Isoleucine131.050.00.3816794
L-Leucine131.075.00.57251906
L-Lysine hydrochloride183.0630.03.442623
L-Methionine149.050.00.33557048
L-Phenylalanine165.0150.00.90909094
L-Proline115.0350.03.0434783
L-Serine105.0550.05.2380953
L-Threonine119.0180.01.5126051
L-Tryptophan204.0100.00.49019608
L-Tyrosine181.055.00.3038674
L-Valine117.0100.00.85470086
Vitamins
Biotin244.00.014.0983607E-5
D-Calcium pantothenate477.00.112.3060797E-4
Folic Acid441.00.024.5351473E-5
Nicotinic acid (Niacin)123.00.021.6260162E-4
Para-Aminobenzoic Acid137.00.021.459854E-4
Pyridoxine hydrochloride206.00.029.708737E-5
Riboflavin376.00.025.319149E-5
Thiamine hydrochloride337.00.025.934718E-5
Vitamin B121355.00.017.380074E-6
i-Inositol180.00.021.11111105E-4
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0980.08.828829
Magnesium Chloride (anhydrous)95.01070.011.263158
Magnesium Sulfate (MgSO4) (anhyd.)120.01370.011.416667
Potassium Chloride (KCl)75.02900.038.666668
Sodium Bicarbonate (NaHCO3)84.0350.04.1666665
Sodium Phosphate dibasic (Na2HPO4-H2O)142.01100.07.746479
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Tryptose Broth2600.0Infinity
参照
Related Products:
43000 - TC 100 Medium, powder
Cat No: 43000033