Hilton Chicago | Chicago, IL
Thermo Fisher Scientific was a Blossom sponsor at CiderCon 2023. At our booth, we discussed our latest applications and workflows for cider analysis, hosted a demonstration of beverage analysis using Gallery discrete analyzer, and our experts answered questions.
To achieve consistent quality and taste for cider and beer, laboratories must analyze multiple parameters—often requiring multiple automated instruments. Yet while each instrument offers precise and accurate results on its own, the entire process is not as fast or automated as it could be, which can impact your efficiency and throughput. Why operate multiple analyzers, when you can do all your essential testing with one? With the liberating technology of Thermo Scientific Gallery discrete analyzers you can deliver reliable analyses for multiple parameters—like bitterness, acidity and sugar content—all from a single sample. This system’s automated workflow and fast analysis work together to ensure high throughput and low cost-per-test. And because the instrument operates without manual intervention, your staff can enjoy walkaway productivity and focus on more strategic tasks in the lab. More testing, from fewer instruments and with fewer techs.
Fill out our form below to speak with an expert. You can also join our community for invitations to our events and information about the solutions we offer.
 Automate your wet chemical analysis
Thermo Scientific Gallery discrete analyzers and Thermo Scientific Gallery system ready-to-use reagents and assay kits. The Thermo Scientific Gallery discrete analyzers can help growing laboratories perform food and beverage analysis, and environmental water analysis by automating labor-intensive and time-consuming multiparameter wet chemical analysis. The high throughput Gallery platform together with ready-to-use system reagents offers the highest flexibility with walkaway efficiency
 Gallery discrete analyzer system for cider analysis
Gallery and Gallery Plus discrete analyzers automate manual wet chemistry methods, photometric (colorimetric and enzymatic), and electrochemical (pH and conductivity) analysis, mimicking the operation of lab chemists, to provide fast, reproducible results in a compact, benchtop design. The discrete cell technology allows laboratories to measure multiple analytes simultaneously while reducing total analysis and operator time. Gallery discrete analyzer, with miniaturized components and unique low-volume cuvette design, accommodates small reagent volumes, minimizes reagent waste, and as a result, reduces analysis cost.
Beverage Testing, your brand is everything
And you can trust us to help keep it that way. You know the power of your brand. You also know your brand’s promise is in every drop. That’s why each of the world’s top ten beverage companies trust us to help keep their products and brands safe, consistent, unadulterated. So they come to us and our widest instrumentation portfolio on the planet—ion, liquid and gas chromatography, metal analysis, mass spectrometry, discrete analyzers, laboratory equipment and plasticware, and data management—for accurate, reliable answers, no matter where they are in the beverage analysis process.
* Required field
Event date white text
CMD Wide-format style fixes
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
CMD Form Code v3.0