Technical Resources - Media Formulations
Gibco® BG-11 Media: Optimized for Cyanobacteria
Gibco® BG-11 Media is optimized for the growth and maintenance of select Cyanobacteria including Synechococcus elongates.
The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps. Each Gibco® BG-11 Media comes in the award winning Gibco® bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helps you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Cyanobacteria culture, results in better overall efficiency and more robust data For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Boric Acid (H3BO3)61.832.860.04625586
Calcium Chloride (CaCl2-2H2O)147.027.00.18367347
Cupric sulfate (CuSO4-5H2O)249.690.0793.1639234E-4
Ferric ammonium citrate261.9812.00.045805022
Magnesium Sulfate (MgSO4-7H2O)246.4775.00.30429667
Manganese chloride (MnCl2-4H2O)197.91.810.009146034
Potassium phosphate dibasic (K2HPO4)174.239.00.2238806
Sodium Molybdate (Na2MoO4-2H20)205.920.3910.0018987957
Sodium Nitrate NaNO384.991500.017.649136
Sodium carbonate (Na2CO3 H2O)105.9920.00.18869705
Zinc sulfate (ZnSO4-7H2O)287.550.2227.720397E-4
Other Components
EDTA292.241.00.0034218451
Verweis
Related Products:
Terrific Broth
Cat No: 22711022
2-YT Broth
Cat No: 22712020
TAP Growth Media, optimized for Chlamydomonas culture
Cat No: A1379801 , A1379802