          • Proteins & Peptides ›
          • Activin A Proteins
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          Human Activin A Recombinant Protein
          View all (9) Activin A proteins
          Tech Support
          Cite Human Activin A Recombinant Protein
          Additional Information:
          {{banner.description}}
          • Testing Data (1)
          Human Activin A Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human Activin A Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Human Activin A Protein (14-8993-62) in Bioactivity
          Inhibition of MPC-11 proliferation by Human Activin A Recombinant Protein. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human Activin A Protein in Bioactivity (Bioactivity)
          Product Details
          14-8993-62
          Product Specifications
          Expression System
          Insect cells
          Class
          Recombinant
          Type
          Protein
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          5 µg
          Storage conditions
          -80°C
          Shipping conditions
          Dry ice
          Product Specific Information
          Description: Activin A is a member of the TGF-beta superfamily, along with the other activin proteins and the structurally related inhibins. The activins are dimeric proteins formed by the association of two of the four existing beta subunits, betaA, betaB, betaC, and betaE. These subunits are shared with the inhibin proteins, which are heterodimers of one beta and one alpha chain. Activin A is a homodimer of two 13 kDa betaA subunits, which can also associate with betaB or betaC to form Activin AB or AC, respectively. Activin A is expressed by various cell types and exhibits pleiotropic effects including the regulation of metabolism, homeostasis, cell proliferation and differentiation, apoptosis, and tissue healing. Activin A has been recently reported to work in synergy with TGF-beta to promote the induction of antigen specific iTreg cells leading to inhibition of Th1 and Th2-mediated responses.
          Applications Reported: Human Activin A Recombinant Protein is biologically active.
          Applications Tested: The ED50 of this protein, as measured by inhibition of MPC-11 mouse plasmacytoma cell proliferation, is 0.75-3 ng/mL. This corresponds to a specific activity of 1.3 x 10e6 - 3.3 x 10e5 Units/mg.
          Source: Insect expressed amino acids Gly311-Ser426, accession number NM_002192.
          Bioactivity: The ED50 of this protein, as measured by inhibition of MPC-11 mouse plasmacytoma cell proliferation, is 0.75-3 ng/mL. This corresponds to a specific activity of 1.3 x 10e6 - 3.3 x 10e5 Units/mg.
          Endotoxin: Less than 0.01 ng/ug cytokine as determined by the LAL assay. Purity: Greater than or equal to 95% as determined by SDS-PAGE.
          Molecular Weight: 26 kDa.
          Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment.
          Purity: Greater than 90%, as determined by SDS-PAGE.
          Aggregation: Less than 10%, as determined by HPLC.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline