          • Primary Antibodies ›
          • ZHX3 Antibodies
          NeoBiotechnologies
          ZHX3 (Transcriptional Repressor) Monoclonal Antibody (PCRP-ZHX3-1G3)
          View all (13) ZHX3 antibodies
          Tech Support
          Cite ZHX3 (Transcriptional Repressor) Monoclonal Antibody (PCRP-ZHX3-1G3)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          ZHX3 (Transcriptional Repressor) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ZHX3 (Transcriptional Repressor) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          ZHX3 (Transcriptional Repressor) Antibody (23051-MSM3-P1) in ICC/IF
          Immunofluorescence analysis of PFA-fixed HeLa cells stained using ZHX3 Mouse Monoclonal Antibody (PCRP-ZHX3-1G3) followed by goat anti-mouse IgG-CF488 (green). CF640A phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZHX3 (Transcriptional Repressor) Antibody in Immunocytochemistry (ICC/IF)
          ZHX3 (Transcriptional Repressor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZHX3 (Transcriptional Repressor) Antibody in Flow Cytometry (Flow)
          ZHX3 (Transcriptional Repressor) Antibody in Peptide array (ARRAY)
          ZHX3 (Transcriptional Repressor) Antibody in SDS-PAGE (SDS-PAGE)
          ZHX3 (Transcriptional Repressor) Monoclonal Antibody (PCRP-ZHX3-1G3)
          Product Details
          23051-MSM3-P1
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          2 µg/mL
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1-2 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          1-2 µg/mL
          -
          Flow Cytometry (Flow)
          1-2 µg/10^6 cells
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          1-2 µg/100-500 µg total protein
          -
          Peptide Array (Array)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          PCRP-ZHX3-1G3
          Immunogen
          Recombinant full-length human ZHX3 protein
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor.
          Bioinformatics
          Protein Aliases: triple homeobox 1; Triple homeobox protein 1; unnamed protein product; Zinc finger and homeodomain protein 3; Zinc fingers and homeoboxes protein 3
          View more View less
          Gene Aliases: KIAA0395; TIX1; ZHX3
          View more View less
          UniProt ID: (Human) Q9H4I2
          View more View less
          Entrez Gene ID: (Human) 23051
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding protein homodimerization activity metal ion binding protein heterodimerization activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription from RNA polymerase II promoter cell differentiation positive regulation of osteoblast differentiation negative regulation of transcription, DNA-templated
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline