    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Mouse IgG2a Secondary Antibodies
          Invitrogen
          Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody
          View all (31) Goat anti-Mouse IgG2a antibodies
          Questions & Answers
          Cite Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody
          Additional Information:
          {{banner.description}}
          Product Details
          31237
          Applications
          Tested Dilution
          Western Blot (WB)
          Assay-dependent
          Immunohistochemistry (IHC)
          Assay-dependent
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          Flow Cytometry (Flow)
          Assay-dependent
          Immunoprecipitation (IP)
          Assay-dependent
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Goat / IgG
          Class
          Polyclonal
          Type
          Secondary Antibody
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.3 mg/mL
          Amount
          1 mg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.6
          Contains
          no preservative
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_429689
          Target
          IgG2a
          Cross Adsorption
          Against human, bovine and rabbit serum proteins
          Antibody Form
          Whole Antibody
          Product Specific Information
          Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration.
          Product # 31237 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications.
          Product # 31237 reacts with the heavy chains of mouse IgG2a, but not with the heavy chains of other mouse IgG subclasses or with light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, this antibody may cross-react with immunoglobulins from other species.
          Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C.
          Country of Origin: USA
          Target Information
          Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody 31237 on CiteAb
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline