    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • VPS4B Antibodies
          Proteintech
          VPS4B Monoclonal Antibody (2H3G7)
          View all (8) VPS4B antibodies
          Tech Support
          Cite VPS4B Monoclonal Antibody (2H3G7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          VPS4B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VPS4B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          VPS4B Antibody (68443-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using VPS4B antibody (68443-1-Ig, Clone: 2H3G7) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          VPS4B Antibody in Immunocytochemistry (ICC/IF)
          VPS4B Antibody in Western Blot (WB)
          VPS4B Antibody in Western Blot (WB)
          VPS4B Antibody in Immunoprecipitation (IP)
          VPS4B Monoclonal Antibody (2H3G7)
          Product Details
          68443-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          0.5-4.0 µg for 1.0-3.0 mg of total protein lysate
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2H3G7
          Immunogen
          VPS4B fusion protein Ag11610
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MSSTSPNLQK AIDLASKAAQ EDKAGNYEEA LQLYQHAVQY FLHVVKYEAQ GDKAKQSIRA KCTEYLDRAE KLKEYLKNKE KKAQKPVKEG QPSPADEKGN DSDGEGESDD PEKKKLQNQL QGAIVIERPN VKWSDVAGLE GAKEALKEAV ILPIKFPHLF TGKRTPWRGI LLFGPPGTGK SYLAKAVATE ANNSTFFSIS SSDLVSKWLG ESEKLVKNLF QLARENKPSI IFIDEIDSLC GSRSENESEA ARRIKTEFLV QMQGVGVDND GILVLGATNI PWVLDSAIRR RFEKRIYIPL PEPHARAAMF KLHLGTTQNS LTEADFRELG RKTDGYSGAD ISIIVRDALM QPVRKVQSAT HFKKVRGPSR ADPNHLVDDL LTPCSPGDPG AIEMTWMDVP GDKLLEPVVS MSDMLRSLSN TKPTVNEHDL LKLKKFTEDF GQEG
          Target Information
          The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. Mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be a yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 18; the gene for the other resides on chromosome 16.
          Bioinformatics
          Protein Aliases: cell migration-inducing 1; Cell migration-inducing gene 1 protein; MIG1; Protein SKD1; Suppressor of K(+) transport growth defect 1; suppressor of K+ transport defect 1; unnamed protein product; vacuolar protein sorting 4B; Vacuolar protein sorting-associated protein 4B
          View more View less
          Gene Aliases: 8030489C12Rik; MIG1; SKD1; SKD1B; VPS4-2; VPS42; VPS4B
          View more View less
          UniProt ID: (Human) O75351, (Mouse) P46467
          View more View less
          Entrez Gene ID: (Human) 9525, (Mouse) 20479, (Rat) 360834
          View more View less
          Function(s)
          nucleotide binding protein binding ATP binding hydrolase activity ATPase activity identical protein binding protein homodimerization activity macromolecular complex binding protein C-terminus binding ATPase activity, coupled molecular_function microtubule-severing ATPase activity non-motor microtubule binding protein
          Process(es)
          angiogenesis plasma membrane repair potassium ion transport autophagy nucleus organization endosome organization vacuole organization vacuolar transport lysosomal transport mitotic metaphase plate congression central nervous system development regulation of G2/M transition of mitotic cell cycle regulation of centrosome duplication positive regulation of G2/M transition of mitotic cell cycle protein transport endosomal transport macroautophagy cholesterol transport nuclear envelope reassembly endosome to lysosome transport via multivesicular body sorting pathway response to lipid multivesicular body assembly viral budding via host ESCRT complex ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway viral budding from plasma membrane protein depolymerization cell division canonical Wnt signaling pathway establishment of blood-brain barrier late endosomal microautophagy late endosome to lysosome transport via multivesicular body sorting pathway midbody abscission multivesicular body sorting pathway membrane fission ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway autophagosome maturation regulation of mitotic spindle assembly negative regulation of exosomal secretion positive regulation of exosomal secretion positive regulation of centriole elongation ESCRT III complex disassembly cell separation after cytokinesis transport cell cycle viral release from host cell positive regulation of viral process regulation of viral process negative regulation of cell death positive regulation of viral release from host cell positive regulation of viral life cycle biological_process cytoplasmic microtubule organization intracellular cholesterol transport
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline