          • Primary Antibodies ›
          • CDY1/2 Antibodies
          Antibodies Inc
          CDY1/2 Monoclonal Antibody (N143/38), FL550
          View all (5) CDY1/2 antibodies
          Tech Support
          Cite CDY1/2 Monoclonal Antibody (N143/38), FL550
          Additional Information:
          {{banner.description}}
          CDY1/2 Monoclonal Antibody (N143/38), FL550
          Product Details
          75-174-FL550
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N143/38
          Immunogen
          Fusion protein amino acids 6-66 (Chromo domain) of human CDY1 (accession number Q9Y6F8) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          550/575 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          CDY1 and CDY2 (Chromodomain Y-linked 1 and 2) are genes located on the Y chromosome that encode proteins involved in chromatin remodeling and spermatogenesis. These proteins contain a chromodomain, which is important for binding to specific regions of chromatin and regulating gene expression during the development of sperm cells. Mutations or deletions in these genes can lead to male infertility due to defects in sperm development.
          Bioinformatics
          Protein Aliases: CDY1; CDY2; chromodomain protein, Y chromosome, 1; chromodomain protein, Y chromosome, 2; chromodomain protein, Y-linked, 1; chromodomain protein, Y-linked, 2A; testis-specific chromodomain protein on Y; Testis-specific chromodomain protein Y 1; Testis-specific chromodomain protein Y 2; unnamed protein product; Y chromosome chromodomain protein 2A
          View more View less
          Gene Aliases: CDY; CDY1; CDY1A; CDY1B; CDY2; CDY2A; CDY2B
          View more View less
          UniProt ID: (Human) Q9Y6F8, (Human) Q9Y6F7
          View more View less
          Entrez Gene ID: (Human) 9085, (Human) 9426
          View more View less
          Function(s)
          transcription corepressor activity histone acetyltransferase activity transferase activity transferase activity, transferring acyl groups H3K27me3 modified histone binding peptide-lysine-N-acetyltransferase activity histone H3K9me2/3 reader activity
          Process(es)
          chromatin organization chromatin remodeling spermatogenesis negative regulation of transcription, DNA-templated
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline