    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • SAPAP1/2 Antibodies
          Antibodies Inc
          SAPAP1/2 Monoclonal Antibody (N238/31), FL650
          View all (5) SAPAP1/2 antibodies
          Tech Support
          Cite SAPAP1/2 Monoclonal Antibody (N238/31), FL650
          Additional Information:
          {{banner.description}}
          SAPAP1/2 Monoclonal Antibody (N238/31), FL650
          Product Details
          75-236-FL650
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N238/31
          Immunogen
          Fusion protein amino acids 772-992 (C-terminus) of rat SAPAP1 (accession number P97836) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          SAPAP1 and SAPAP2 refer to Synapse-Associated Protein 90/Postsynaptic Density-95-Associated Protein 1 and 2, respectively. These proteins are part of a family of scaffolding proteins that play a crucial role in the organization and function of synapses in the brain. SAPAP1 and SAPAP2 are important scaffolding proteins that contribute to the structural and functional organization of synapses, playing a key role in synaptic signaling and plasticity.
          Bioinformatics
          Protein Aliases: DAP-1; DAP-1 beta; DAP-2; discs large homolog associated protein 1; discs large homolog associated protein 2; discs large homolog-associated protein 2; discs large-associated protein 2; Disks large-associated protein 1; Disks large-associated protein 2; ERICH1 antisense gene protein 1; ERICH1 antisense RNA 1 (non-protein coding); guanylate kinase associated protein; Guanylate kinase-associated protein; hGKAP; postsynaptic density 95; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-associated protein-2; PSD-95/SAP90-binding protein 1; PSD-95/SAP90-binding protein 2; putative DAP-2 like protein; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95 associated protein 2; SAP90/PSD-95-associated protein 1; SAP90/PSD-95-associated protein 2; SAPAP1; SAPAP2; unnamed protein product
          View more View less
          Gene Aliases: 4933422O14Rik; 6430596N04Rik; 9630002F18; AI845682; AI848168; BB075781; C8orf68; D17Bwg0511e; DAP-1; DAP-1-ALPHA; DAP-1-BETA; DAP-2; DAP1; DAP2; DLGAP1; DLGAP1A; DLGAP1B; Dlgap2; ERICH1-AS1; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; Kiaa4162; mKIAA4162; rGKAP; SAPAP1; SAPAP2
          View more View less
          UniProt ID: (Human) O14490, (Mouse) Q9D415, (Rat) P97836, (Rat) P97837, (Mouse) Q8BJ42
          View more View less
          Entrez Gene ID: (Human) 9229, (Human) 9228, (Mouse) 224997, (Rat) 65040, (Rat) 116681, (Mouse) 244310
          View more View less
          Function(s)
          protein binding macromolecular complex binding binding, bridging protein domain specific binding protein complex binding guanylate kinase activity scaffold/adaptor protein
          Process(es)
          chemical synaptic transmission signaling modulation of synaptic transmission neuron-neuron synaptic transmission protein localization to synapse regulation of proteasomal protein catabolic process aggresome assembly cell-cell signaling nucleotide phosphorylation biological_process
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline