          • Primary Antibodies ›
          • alpha COP Antibodies
          Proteintech
          COPA Recombinant Rabbit Monoclonal Antibody (230361B1)
          View all (9) alpha COP antibodies
          Tech Support
          Cite COPA Recombinant Rabbit Monoclonal Antibody (230361B1)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          COPA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          COPA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          COPA Antibody (83089-2-RR) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using COPA antibody (83089-2-RR, Clone: 230361B1) at dilution of 1:250 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          COPA Antibody in Immunocytochemistry (ICC/IF)
          COPA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          COPA Antibody in Western Blot (WB)
          COPA Antibody in Flow Cytometry (Flow)
          COPA Recombinant Rabbit Monoclonal Antibody (230361B1)
          Product Details
          83089-2-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:125-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230361B1
          Immunogen
          COPA fusion protein Ag34207
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: QLPVDHILAG SFETAMRLLH DQVGVIQFGP YKQLFLQTYA RGRTTYQALP CLPSMYGYPN RNWKDAGLKN GVPAVGLKLN DLIQRLQLCY QLTTVGKFEE AVEKFRSILL SVPLLVVDNK QEIAEAQQLI TICREYIVGL SVETERKKLP KETLEQQKRI CEMAAYFTHS NLQPVHMILV LRTALNLFFK LKNFKTAATF ARRLLELGPK PEVAQQTRKI LSACEKNPTD AYQLNYDMHN PFDICAASYR PIYRGKPVEK CPLSGACYSP EFKGQICRVT TVTEIGKDVI GLRISPLQFR
          Target Information
          In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
          Bioinformatics
          Protein Aliases: alpha coat protein; Alpha-coat protein; Alpha-COP; coatomer protein complex subunit alpha; Coatomer subunit alpha; HEP-COP; HEPCOP; proxenin; xenin
          View more View less
          Gene Aliases: AIAISD; AILJK; alpha-COP; AU040324; COPA; HEP-COP; xenin
          View more View less
          UniProt ID: (Human) P53621, (Mouse) Q8CIE6
          View more View less
          Entrez Gene ID: (Human) 1314, (Mouse) 12847
          View more View less
          Function(s)
          mRNA binding hormone activity structural molecule activity protein binding vesicle coat protein
          Process(es)
          intracellular protein transport ER to Golgi vesicle-mediated transport retrograde vesicle-mediated transport, Golgi to ER intra-Golgi vesicle-mediated transport signal transduction protein transport vesicle-mediated transport pancreatic juice secretion protein localization to axon protein localization to cell leading edge transport
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline