          • Primary Antibodies ›
          • CD163 Antibodies
          NeoBiotechnologies
          CD163 (Monocyte and Macrophage Marker) Monoclonal Antibody (M130/1210)
          View all (140) CD163 antibodies
          Tech Support
          Cite CD163 (Monocyte and Macrophage Marker) Monoclonal Antibody (M130/1210)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          CD163 (Monocyte and Macrophage Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CD163 (Monocyte and Macrophage Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          CD163 (Monocyte and Macrophage Marker) Antibody (9332-MSM1-P1ABX) in IHC (P)
          Formalin-fixed, paraffin-embedded human Histiocytoma stained with CD163 Mouse Monoclonal Antibody (M130/1210). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD163 (Monocyte and Macrophage Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD163 (Monocyte and Macrophage Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD163 (Monocyte and Macrophage Marker) Antibody in Western Blot (WB)
          CD163 (Monocyte and Macrophage Marker) Antibody in Peptide array (ARRAY)
          CD163 (Monocyte and Macrophage Marker) Antibody in SDS-PAGE (SDS-PAGE)
          CD163 (Monocyte and Macrophage Marker) Monoclonal Antibody (M130/1210)
          Product Details
          9332-MSM1-P1ABX
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-dependent
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1-2 µg/mL
          -
          Peptide Array (Array)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          M130/1210
          Immunogen
          Recombinant human CD163 fragment (around aa 43-196).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD163 (M130 antigen, Ber-Mac3, Ki-M8, SM4) is a 130 kDa membrane glycoprotein, a member of the scavenger receptor cysteine-rich superfamily, and a receptor for the hemoglobin-haptoglobin complex. CD163 protects tissues from free hemoglobin-mediated oxidative damage, and may play a role in the uptake and recycling of iron, via endocytosis of hemoglobin/haptoglobin and subsequent breakdown of heme. CD163 is expressed exclusively on the cell surface of human monocytes and macrophages that evolve predominantly in the late phase of inflammation. Specifically, CD163 is present on all circulating monocytes and most tissue macrophages except those found in the mantle zone and germinal centers of lymphoid follicles, interdigitating reticulum cells and Langerhan's cells. CD163 is present on all CD14 positive monocytes, most CD64 positive monocytes, and shows higher expression on CD16 positive monocytes. CD163 is upregulated on mononuclear phagocytes by IL-10, IL-6 and dexamethasone. Lipopolysaccharide (LPS) and phorbol myristate acetate (PMA) both induce shedding of CD163 from the cell surface into plasma or cell supernatant. CD163 binds hemoglobin/haptoglobin complexes in a calcium-dependent and pH-dependent manner, and exhibits a higher affinity for complexes of hemoglobin and multimeric haptoglobin of HP1F phenotype than for complexes of hemoglobin and dimeric haptoglobin of HP1S phenotype. Further, CD163 also induces a cascade of intracellular signals that involves tyrosine kinase-dependent calcium mobilization, inositol triphosphate production and secretion of IL6 and CSF1.
          Bioinformatics
          Protein Aliases: CD163; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; Scavenger receptor cysteine-rich type 1 protein M130; Soluble sCD163
          View more View less
          Gene Aliases: CD163; M130; MM130; SCARI1
          View more View less
          UniProt ID: (Human) Q86VB7
          View more View less
          Entrez Gene ID: (Human) 9332
          View more View less
          Function(s)
          scavenger receptor activity protein binding scaffold protein binding
          Process(es)
          acute-phase response inflammatory response vesicle-mediated transport
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline