          • Primary Antibodies ›
          • DOCK10 Antibodies
          Bethyl Laboratories
          DOCK10 Polyclonal Antibody
          View all (3) DOCK10 antibodies
          Tech Support
          Cite DOCK10 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          DOCK10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DOCK10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          DOCK10 Antibody (A301-305A-T) in WB
          Detection of human DOCK10 by western blot. Samples: Whole cell lysate (50 µg) from Jurkat, LNCaP, and GaMG cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-DOCK10 antibody (Product # A301-305A) (lot A301-305A-2) used for WB at 0.1 µg/mL. Detection: Chemiluminescence with an exposure time of 10 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DOCK10 Antibody in Western Blot (WB)
          DOCK10 Antibody in Immunoprecipitation (IP)
          DOCK10 Polyclonal Antibody
          Product Details
          A301-305A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 100 and 150 of human dedicator of cytokinesis 10.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms.
          Bioinformatics
          Protein Aliases: Dedicator of cytokinesis protein 10; dopamine receptor interacting protein 2; unnamed protein product; Zizimin-3; zizimin3
          View more View less
          Gene Aliases: DOCK10; DRIP2; KIAA0694; Nbla10300; ZIZ3
          View more View less
          UniProt ID: (Human) Q96BY6
          View more View less
          Entrez Gene ID: (Human) 55619
          View more View less
          Function(s)
          guanyl-nucleotide exchange factor activity protein binding small GTPase binding
          Process(es)
          B cell homeostasis marginal zone B cell differentiation small GTPase mediated signal transduction regulation of cell migration regulation of Rho protein signal transduction positive regulation of GTPase activity dendritic spine morphogenesis regulation of postsynapse assembly
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline