          • Primary Antibodies ›
          • RC3H2 Antibodies
          Bethyl Laboratories
          RC3H2/MNAB Polyclonal Antibody
          View all (4) RC3H2 antibodies
          Tech Support
          Cite RC3H2/MNAB Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          RC3H2/MNAB Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          RC3H2/MNAB Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          RC3H2/MNAB Antibody (A305-149A-T) in IP
          Detection of human RC3H2 by western blot of immunoprecipitates. Samples: Whole cell lysate (0.5 or 1.0 mg per IP reaction; 20% of IP loaded) from HeLa cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-RC3H2 antibody (Product # A305-149A) (lot 1) used for IP at 6 µg per reaction. RC3H2 was also immunoprecipitated by rabbit anti-RC3H2 antibody A305-150A. For blotting immunoprecipitated RC3H2, A305-149A was used at 0.4 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RC3H2/MNAB Antibody in Immunoprecipitation (IP)
          RC3H2/MNAB Polyclonal Antibody
          Product Details
          A305-149A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 900 to 950 of Human RING finger and CCCH-type zinc finger domain-containing protein 2.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Mouse
          Target Information
          RC3H2 gene ontology annotations related to this gene include B cell homeostasis; limb development; lung alveolus development; lymph node development; multicellular organism growth; negative regulation of T-helper 17 cell differentiation; positive regulation of NIK/NF-kappaB signaling; post-embryonic development; posttranscriptional regulation of gene expression; regulation of miRNA metabolic process; spleen development; T cell homeostasis; T cell proliferation; T cell receptor signaling pathway; T follicular helper cell differentiation.
          Bioinformatics
          Protein Aliases: FLJ20301; FLJ20713; membrane associated DNA binding protein; Membrane-associated nucleic acid-binding protein; MNAB; RING finger and CCCH-type zinc finger domain-containing protein 2; ring finger and CCCH-type zinc finger domains 2; RING finger protein 164; RING-type E3 ubiquitin transferase Roquin-2; Roquin-2; unnamed protein product
          View more View less
          Gene Aliases: MNAB; RC3H2; RNF164
          View more View less
          UniProt ID: (Human) Q9HBD1
          View more View less
          Entrez Gene ID: (Human) 54542
          View more View less
          Function(s)
          DNA binding RNA binding double-stranded RNA binding mRNA binding zinc ion binding transferase activity RNA stem-loop binding metal ion binding ubiquitin protein ligase activity
          Process(es)
          protein polyubiquitination nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay B cell homeostasis ubiquitin-dependent protein catabolic process post-embryonic development regulation of gene expression posttranscriptional regulation of gene expression protein ubiquitination multicellular organism growth T cell proliferation T cell homeostasis lung alveolus development lymph node development spleen development T cell receptor signaling pathway limb development T follicular helper cell differentiation positive regulation of NIK/NF-kappaB signaling negative regulation of T-helper 17 cell differentiation regulation of miRNA metabolic process
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline