          • Primary Antibodies ›
          • Ki-67 Antibodies
          Bethyl Laboratories
          Ki-67 Recombinant Rabbit Monoclonal Antibody (BLR021E)
          View all (213) Ki-67 antibodies
          Tech Support
          Cite Ki-67 Recombinant Rabbit Monoclonal Antibody (BLR021E)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          Ki-67 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Ki-67 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          Ki-67 Antibody (A700-021-T) in ICC/IF
          Detection of human Ki-67 by immunocytochemistry. Sample: acetone/methanol fixed Hela cells. Antibody: Rabbit anti-Ki-67 recombinant monoclonal antibody [BLR021E] (Product # A700-021) (lot 1) used at 1:100. Secondary: DyLight® 594-conjugated goat anti-rabbit IgG (A120-101D4). Counterstain: DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Ki-67 Antibody in Immunocytochemistry (ICC/IF)
          Ki-67 Antibody in Immunocytochemistry (ICC/IF)
          Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ki-67 Antibody in Flow Cytometry (Flow)
          Ki-67 Recombinant Rabbit Monoclonal Antibody (BLR021E)
          Product Details
          A700-021-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:500
          -
          Flow Cytometry (Flow)
          1 µL per 1 x 10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          BLR021E
          Immunogen
          Peptide representing the repetitive 66 bp element of human Ki-67.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          50 µg/mL
          Amount
          0.5 µg
          Purification
          Ion-exchange chromatography
          Storage buffer
          BBS, pH 8.2, with 0.1% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
          Target Information
          Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with euchromatin at the perichromosomal layer, the centromeric heterochromatin, and the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis. Ki-67 antibodies are useful in establishing the cell growing fraction in neoplasms. In neoplastic tissues, the prognostic value is comparable to the tritiated thymidine-labelling index. The correlation between low Ki-67 index and histologically low-grade tumors is strong. Ki-67 is routinely used as a neuronal marker of cell cycling and proliferation.
          Bioinformatics
          Protein Aliases: Antigen identified by monoclonal antibody Ki-67; Antigen KI-67; antigen Ki67; Molecular Immunology Borstel antibody 1; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; unnamed protein product
          View more View less
          Gene Aliases: KIA; MIB-; MIB-1; MKI67; PPP1R105
          View more View less
          UniProt ID: (Human) P46013
          View more View less
          Entrez Gene ID: (Human) 4288
          View more View less
          Function(s)
          nucleotide binding DNA binding RNA binding protein binding ATP binding molecular condensate scaffold activity
          Process(es)
          chromosome segregation regulation of mitotic nuclear division cell proliferation regulation of chromatin organization
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline