          • Primary Antibodies ›
          • MAFB Antibodies
          Bethyl Laboratories
          MafB Recombinant Rabbit Monoclonal Antibody (BLR046F)
          View all (23) MAFB antibodies
          Tech Support
          Cite MafB Recombinant Rabbit Monoclonal Antibody (BLR046F)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          MafB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MafB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          MafB Antibody (A700-046-T) in ICC/IF
          Detection of human MAFB in FFPE SK-N-MC cells by immunocytochemistry. Antibody: Rabbit anti-MafB recombinant monoclonal antibody [BLR046F] (Product # A700-046) (lot 3). Secondary: HRP-conjugated goat anti-rabbit IgG (A120-501P). Substrate: DAB. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MafB Antibody in Immunocytochemistry (ICC/IF)
          MafB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MafB Antibody in Western Blot (WB)
          MafB Antibody in Western Blot (WB)
          MafB Antibody in Immunoprecipitation (IP)
          MafB Recombinant Rabbit Monoclonal Antibody (BLR046F)
          Product Details
          A700-046-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100 - 1:500
          -
          Immunocytochemistry (ICC/IF)
          1:100 - 1:500
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          BLR046F
          Immunogen
          Peptide representing a region between residues 1 and 50 of human MAF bZIP transcription factor B (MafB).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          10 µg
          Purification
          purified
          Storage buffer
          BBS, pH 7.8 to 8.6
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.
          Bioinformatics
          Protein Aliases: bZIP transcription factor; Kreisler; Kreisler (maf-related) leucine zipper protein; Kreisler maf-related leucine zipper homolog; Maf-B; MAFB/Kreisler basic region/leucine zipper transcription factor; MGC43127; Segmentation protein Kr; Transcription factor Maf-1; Transcription factor MafB; unnamed protein product; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B; V-maf musculoaponeurotic fibrosarcoma oncogene homolog B
          View more View less
          Gene Aliases: DURS3; kr; Kreisler; KRML; Krml1; Maf1; MAFB; MCTO
          View more View less
          UniProt ID: (Human) Q9Y5Q3, (Mouse) P54841
          View more View less
          Entrez Gene ID: (Human) 9935, (Mouse) 16658
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding identical protein binding sequence-specific DNA binding macromolecular complex binding sequence-specific double-stranded DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcription factor binding basic leucine zipper transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter segment specification sensory organ development response to nutrient respiratory gaseous exchange gene expression protein processing rhombomere 5 development rhombomere 6 development abducens nerve formation keratinocyte differentiation T cell differentiation in thymus brain segmentation inner ear morphogenesis fat cell differentiation regulation of myeloid cell differentiation negative regulation of erythrocyte differentiation negative regulation of osteoclast differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter thymus development integrated stress response signaling cornified envelope assembly transcription, DNA-templated transcription from RNA polymerase II promoter
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline