Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MACLMAAFS VGTAMNASSY SAEMTEPKSV CVSVDEVVSS NMEATETDLL NGHLKKVDNN LTEAQRFSSL PRRAAVNIEF RDLSYSVPEG PWWRKKGYKT LLKGISGKFN SGELVAIMGP SGAGKSTLMN ILAGYRETGM KGAVLINGLP RDLRCFRKVS CYIMQDDMLL PHLTVQEAMM VSAHLKLQEK DEGRREMVKE ILTALGLLSC ANTRTGSLSG GQRKRLAIAL ELVNNPPVMF FDEPTSGLDS ASCFQVVSLM KGLAQGGRSI ICTIHQPSAK LFELFDQLYV LSQGQCVYRG KVCNLVPYLR DLGLNCPTYH NPADFVMEVA SGEYGDQNSR LVRAVREGMC DSDH (1-353 aa encoded by BC029158)
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. The ABC transporter ABCG1 (ABC8) is expressed on the cell surface and in intracellular compartments of cholesterol-laden macrophages and is induced in monocyte-derived macrophages during cholesterol influx mediated by acetylated low-density lipoprotein. In contrast, lipid efflux in cholesterol-laden macrophages, mediated by the cholesterol acceptor HDL(3), suppresses the expression of ABCG1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC transporter 8; ABCG1p; ATP-binding cassette 8; ATP-binding cassette transporter 8; ATP-binding cassette transporter 8 (ABC8); ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette transporter, member 1 of subfamily G; ATP-binding cassette, sub-family G (WHITE), member 1; ATP-binding cassette, subfamily G (WHITE), member 1; homolog of Drosophila white; MGC34313; unnamed protein product; white; white protein homolog; white protein homolog (ATP-binding cassette transporter 8)
Gene Aliases: ABC8; White; WHITE1
UniProt ID: (Human) P45844, (Mouse) Q64343
Entrez Gene ID: (Human) 9619, (Rat) 85264, (Mouse) 11307
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support