          • Primary Antibodies ›
          • CCR9 Antibodies
          Alomone Labs, LTD
          CCR9 (extracellular) Polyclonal Antibody
          View all (27) CCR9 antibodies
          Tech Support
          Cite CCR9 (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CCR9 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CCR9 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CCR9 (extracellular) Antibody (ACR-029-50UL) in WB
          Western blot analysis of human acute T cell leukemia (Jurkat) (lanes 1 and 3), human melanoma (MALME-3M) (lanes 2 and 4), human adenocarcinoma (HT-29) (lanes 5 and 7) and mouse T-cell lymphoma (TK-1) (lanes 6 and 8) cell line lysates: - 1,2,5,6. Anti-CCR9 (extracellular) Antibody (#ACR-029), (1:200).3,4,7,8. Anti-CCR9 (extracellular) Antibody , preincubated with CCR9 (extracellular) Blocking Peptide (#BLP-C... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CCR9 (extracellular) Antibody in Western Blot (WB)
          CCR9 (extracellular) Antibody in Flow Cytometry (Flow)
          CCR9 (extracellular) Polyclonal Antibody
          Product Details
          ACR-029-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Flow Cytometry (Flow)
          5 µg
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)DESTKLKSAVLTLK, corresponding to amino acid residues 205-218 of human CCR9, 2nd extracellular loop
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described.
          Bioinformatics
          Protein Aliases: C-C chemokine receptor type 9; C-C CKR-9; CC-CKR-9; CCR-9; CCR9A; CCR9B; CDw199; CDw199 antigen; chemokine (C-C motif) receptor 9; Chemokine C-C receptor 10; G protein-coupled receptor; G protein-coupled receptor 28; G-protein coupled receptor 28; GPR-9-6; unnamed protein product
          View more View less
          Gene Aliases: A130091K22Rik; CC-CKR-9; CCR9; CDw199; Cmkbr10; GPR-9-6; GPR28
          View more View less
          UniProt ID: (Human) P51686, (Mouse) Q9WUT7
          View more View less
          Entrez Gene ID: (Human) 10803, (Rat) 282832, (Mouse) 12769
          View more View less
          Function(s)
          G-protein coupled receptor activity chemokine receptor activity C-C chemokine receptor activity C-C chemokine binding signal transducer activity protein binding
          Process(es)
          chemotaxis immune response G-protein coupled receptor signaling pathway chemokine-mediated signaling pathway signal transduction CD8-positive, gamma-delta intraepithelial T cell differentiation cellular defense response positive regulation of cytosolic calcium ion concentration calcium-mediated signaling cell chemotaxis
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline