          • Primary Antibodies ›
          • Endothelin B Receptor Antibodies
          Alomone Labs, LTD
          Endothelin Receptor B Polyclonal Antibody
          View all (17) Endothelin B Receptor antibodies
          Tech Support
          Cite Endothelin Receptor B Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          Endothelin Receptor B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Endothelin Receptor B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Endothelin Receptor B Antibody (AER-002-50UL) in IHC (P)
          Expression of EDNRB in rat lung - Immunohistochemical staining of paraffin embedded section of rat lung using Anti-Endothelin Receptor B Antibody (#AER-002) (1:100). EDNRB is expressed both in respiratory epithelium (black arrows) and in respiratory smooth muscle (red arrows). Note that vascular smooth muscle is not stained (blue arrows).Hematoxilin is used as the counterstain. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Endothelin Receptor B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endothelin Receptor B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Endothelin Receptor B Antibody in Western Blot (WB)
          Endothelin Receptor B Polyclonal Antibody
          Product Details
          AER-002-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          CEMLRKKSGMQIALND, corresponding to amino acid residues 298-314 of rat ET-B, 3rd intracellular loop
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          50 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
          Bioinformatics
          Protein Aliases: AU022549; endothelin B receptor; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; endothelin-B receptor; ET-B; Et-b receptor; Et-b-r; ET-BR; ET-RB; Etb receptor; Etb-svr; G-protein coupled receptor; Hirschsprung disease 2; piebald; RP11-318G21.1; unnamed protein product
          View more View less
          Gene Aliases: ABCDS; Ednra; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETR-b; ETRB; HSCR; HSCR2; Sox10m1; WS4A
          View more View less
          UniProt ID: (Human) P24530, (Rat) P21451, (Mouse) P48302
          View more View less
          Entrez Gene ID: (Human) 1910, (Rat) 50672, (Mouse) 13618
          View more View less
          Function(s)
          G-protein coupled receptor activity endothelin receptor activity protein binding peptide hormone binding type 1 angiotensin receptor binding receptor activity signal transducer activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter neural crest cell migration renin secretion into blood stream regulation of heart rate regulation of pH signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway negative regulation of adenylate cyclase activity phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration nervous system development peripheral nervous system development posterior midgut development regulation of blood pressure positive regulation of cell proliferation gene expression negative regulation of neuron maturation vein smooth muscle contraction vascular smooth muscle contraction calcium-mediated signaling cGMP-mediated signaling heparin metabolic process melanocyte differentiation regulation of fever generation aldosterone metabolic process response to lipopolysaccharide enteric smooth muscle cell differentiation positive regulation of urine volume renal sodium excretion epithelial fluid transport vasoconstriction vasodilation negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling pigmentation developmental pigmentation macrophage chemotaxis response to pain enteric nervous system development regulation of epithelial cell proliferation negative regulation of protein metabolic process sodium ion homeostasis canonical Wnt signaling pathway positive regulation of penile erection establishment of endothelial barrier renal sodium ion absorption calcium ion transmembrane transport cellular response to lipopolysaccharide protein transmembrane transport glomerular visceral epithelial cell differentiation endothelin receptor signaling pathway renal albumin absorption neuroblast migration chordate pharynx development response to sodium phosphate response to endothelin positive regulation of protein phosphorylation aging response to organic cyclic compound sensory perception of pain negative regulation of cellular protein metabolic process positive regulation of renal sodium excretion regulation of sensory perception of pain
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline