          • Primary Antibodies ›
          • KCNK12 Antibodies
          Alomone Labs, LTD
          KCNK12 (THIK-2) (extracellular) Polyclonal Antibody
          View all (9) KCNK12 antibodies
          Tech Support
          Cite KCNK12 (THIK-2) (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          KCNK12 (THIK-2) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          KCNK12 (THIK-2) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          KCNK12 (THIK-2) (extracellular) Antibody (APC-169-50UL) in IHC (F)
          Expression of KCNK12 (THIK-2) in rat hippocampus - Immunohistochemical staining of perfusion-fixed frozen rat brain sections with Anti-KCNK12 (THIK-2) (extracellular) Antibody (#APC-169), (1:400), followed by goat Anti-rabbit-AlexaFluor-488 secondary Antibody . THIK-2 immunoreactivity (green) in the rat hippocampal CA3 region is detected in neuronal soma (horizontal arrows) and in the dendrite initial segme... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KCNK12 (THIK-2) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          KCNK12 (THIK-2) (extracellular) Antibody in Western Blot (WB)
          KCNK12 (THIK-2) (extracellular) Antibody in Western Blot (WB)
          KCNK12 (THIK-2) (extracellular) Antibody in Western Blot (WB)
          KCNK12 (THIK-2) (extracellular) Polyclonal Antibody
          Product Details
          APC-169-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Frozen) (IHC (F))
          1:400
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)ESPGEAEARARWG, corresponding to amino acid residues 61-73 of rat KCNK12, 1st extracellular loop
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].
          Bioinformatics
          Protein Aliases: Potassium channel subfamily K member 12; potassium channel, subfamily K, member 12; potassium channel, two pore domain subfamily K, member 12; Tandem pore domain halothane-inhibited potassium channel 2; tandem pore domain potassium channel THIK-2; THIK-2
          View more View less
          Gene Aliases: K2p12.1; KCNK12; mntk1; THIK-2; THIK2
          View more View less
          UniProt ID: (Human) Q9HB15, (Rat) Q9ERS1
          View more View less
          Entrez Gene ID: (Human) 56660, (Rat) 64119, (Mouse) 210741
          View more View less
          Function(s)
          potassium channel activity protein binding outward rectifier potassium channel activity potassium ion leak channel activity identical protein binding metal ion binding protein heterodimerization activity voltage-gated ion channel activity molecular_function ion channel transporter
          Process(es)
          ion transport potassium ion transport ion transmembrane transport potassium ion transmembrane transport regulation of ion transmembrane transport biological_process stabilization of membrane potential
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline