Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Amylase is an enzyme that catalyses the breakdown of starch into sugars. Amylase is present in human saliva, where it begins the chemical process of digestion. By in situ hybridization combined with high resolution cytogenetics, the amylase gene is mapped to 1p21. Amylase enzymes find use in bread making and to break down complex sugars such as starch (found in flour) into simple sugars. Yeast then feeds on these simple sugars and converts it into the waste products of alcohol and CO2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 1; alpha amylase 1; Alpha-amylase 1; Alpha-amylase 1A; Alpha-amylase 1B; Alpha-amylase 1C; AMY2A; amylase 1, salivary; amylase, salivary, alpha-1A; glycogenase; Salivary alpha-amylase; salivary amylase; salivary amylase alpha 1A; Salivary and hepatic alpha-amylase
Gene Aliases: Amy-1; Amy-1-a; AMY1; AMY1A; AMY1B; AMY1C; C030014B17Rik
UniProt ID: (Human) P0DUB6, (Mouse) P00687
Entrez Gene ID: (Human) 276, (Mouse) 11722, (Rat) 24203
Molecular Function: amylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support