    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD105 (Endoglin) Antibodies
          OriGene
          ENG Monoclonal Antibody (OTI3D5), TrueMAB™
          View all (147) CD105 (Endoglin) antibodies
          Tech Support
          Cite ENG Monoclonal Antibody (OTI3D5), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          ENG Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ENG Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ENG Antibody (CF506167) in ICC/IF
          Immunofluorescent staining of HeLa cells using anti-ENG mouse monoclonal antibody (TA506167). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ENG Antibody in Immunocytochemistry (ICC/IF)
          ENG Antibody in Western Blot (WB)
          ENG Monoclonal Antibody (OTI3D5), TrueMAB™
          Product Details
          CF506167
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:4,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI3D5
          Immunogen
          Full length human recombinant protein of human ENG produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • Request custom conjugation
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 8% trehalose
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
          Target Information
          CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
          Bioinformatics
          Protein Aliases: CD105; CD105 antigen; circulating isoform of endoglin TGF-beta receptor type III; Endoglin; endoglin precursor; ORW; soluble endoglin; unnamed protein product
          View more View less
          Gene Aliases: END; ENG; HHT1; ORW1
          View more View less
          UniProt ID: (Human) P17813
          View more View less
          Entrez Gene ID: (Human) 2022
          View more View less
          Function(s)
          transmembrane signaling receptor activity type II transforming growth factor beta receptor binding protein binding galactose binding glycosaminoglycan binding coreceptor activity receptor activator activity type I transforming growth factor beta receptor binding BMP binding identical protein binding protein homodimerization activity activin binding transforming growth factor beta binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis patterning of blood vessels vasculogenesis response to hypoxia negative regulation of endothelial cell proliferation heart looping sprouting angiogenesis positive regulation of systemic arterial blood pressure outflow tract septum morphogenesis epithelial to mesenchymal transition involved in endocardial cushion formation endocardial cushion morphogenesis cardiac ventricle morphogenesis cardiac atrium morphogenesis ventricular trabecula myocardium morphogenesis cell migration involved in endocardial cushion formation regulation of transcription, DNA-templated cell adhesion transforming growth factor beta receptor signaling pathway heart development negative regulation of gene expression regulation of cardiac muscle cell apoptotic process cell migration regulation of transforming growth factor beta receptor signaling pathway central nervous system vasculogenesis extracellular matrix disassembly regulation of cell adhesion negative regulation of cell migration BMP signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of BMP signaling pathway dorsal aorta morphogenesis wound healing regulation of cell proliferation positive regulation of angiogenesis positive regulation of transcription from RNA polymerase II promoter smooth muscle tissue development artery morphogenesis venous blood vessel morphogenesis cell motility positive regulation of protein kinase B signaling atrial cardiac muscle tissue morphogenesis cell chemotaxis positive regulation of SMAD protein import into nucleus negative regulation of SMAD protein import into nucleus detection of hypoxia endocardial cushion to mesenchymal transition vascular smooth muscle cell development positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation positive regulation of vascular smooth muscle cell differentiation atrioventricular canal morphogenesis regulation of cardiac neural crest cell migration involved in outflow tract morphogenesis regulation of cell proliferation involved in heart morphogenesis
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0