          • Primary Antibodies ›
          • FAM114A1 Antibodies
          Proteintech
          FAM114A1 Monoclonal Antibody (1B11D6), CoraLite® Plus 488
          View all (8) FAM114A1 antibodies
          Tech Support
          Cite FAM114A1 Monoclonal Antibody (1B11D6), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          FAM114A1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FAM114A1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          FAM114A1 Antibody (CL488-67926) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 FAM114A1 antibody (CL488-67926, Clone: 1B11D6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FAM114A1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-67926
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1B11D6
          Immunogen
          FAM114A1 fusion protein Ag17289
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LEILSNESE SKVQSFLASL DGEKLELLKN DLISIKDIFA AKELENEENQ EEQGLEEKGE EFARMLTELL FELHVAATPD KLNKAMKRAH DWVEEDQTVV SVDVAKVSEE ETKKEEKEEK SQDPQEDKKE EKKTKTIEEV YMSSIESLAE VTARCIEQLH KVAELILHGQ EEEKPAQDQA KVLIKLTTAM CNEVASLSKK FTNSLTTVGS NKKAEVLNPM ISSVLLEGCN STTYI
          Target Information
          May play a role in neuronal cell development.
          Bioinformatics
          Protein Aliases: family with sequence similarity 114, member A1; nervous system over-expressed protein 20; Nervous system overexpressed protein 20; Protein FAM114A1; Protein NOXP20; unnamed protein product
          View more View less
          Gene Aliases: 1190001N04Rik; 9130005N14Rik; FAM114A1; NOXP20; RGD1560201
          View more View less
          UniProt ID: (Human) Q8IWE2, (Mouse) Q9D281
          View more View less
          Entrez Gene ID: (Human) 92689, (Rat) 498366, (Mouse) 68303
          View more View less
          Function(s)
          protein binding molecular_function
          Process(es)
          heart valve morphogenesis inflammatory response gene expression fibroblast migration angiotensin-activated signaling pathway fibroblast proliferation tissue remodeling fibroblast activation connective tissue replacement biological_process
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline