          • Primary Antibodies ›
          • STRA8 Antibodies
          Proteintech
          STRA8 Monoclonal Antibody (1D7E3), CoraLite® Plus 488
          View all (7) STRA8 antibodies
          Tech Support
          Cite STRA8 Monoclonal Antibody (1D7E3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          STRA8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          STRA8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          STRA8 Antibody (CL488-68071) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed NCCIT cells using CoraLite® Plus 488 STRA8 antibody (CL488-68071, Clone: 1D7E3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          STRA8 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68071
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rabbit
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D7E3
          Immunogen
          STRA8 fusion protein Ag29680
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: NLSEERKAS LRQAWAQKHR GPATLAEACR EPACAEGSVK DSGVDSQGAS CSLVSTPEEI LFEDAFDVAS FLDKSEVPST SSSSSVLASC NPENPEEKFQ LYMQIINFFK GLSCANTQVK QEASFPVDEE MIMLQCTETF DDEDL
          Target Information
          Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.
          Bioinformatics
          Protein Aliases: Stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8
          View more View less
          Gene Aliases: STRA8
          View more View less
          UniProt ID: (Human) Q7Z7C7, (Mouse) P70278
          View more View less
          Entrez Gene ID: (Human) 346673, (Rabbit) 103348647, (Mouse) 20899
          View more View less
          Function(s)
          protein binding protein dimerization activity
          Process(es)
          ovarian follicle development DNA replication female meiosis sister chromatid cohesion synapsis reciprocal meiotic recombination spermatogenesis fertilization meiotic chromosome condensation meiotic DNA replication checkpoint meiotic DNA double-strand break formation regulation of organ growth oocyte development meiotic cell cycle cellular response to retinoic acid regulation of transcription from RNA polymerase II promoter cell differentiation oogenesis activation of meiosis
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline