          • Primary Antibodies ›
          • UBE2D1/2/3/4 Antibodies
          Proteintech
          UBE2D1/2/3/4 Monoclonal Antibody (1C7D12), CoraLite® Plus 488
          Tech Support
          Cite UBE2D1/2/3/4 Monoclonal Antibody (1C7D12), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          UBE2D1/2/3/4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UBE2D1/2/3/4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          UBE2D1/2/3/4 Antibody (CL488-68138) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed A431 cells using CoraLite® Plus 488 UBE2D1/2/3/4 antibody (CL488-68138, Clone: 1C7D12) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UBE2D1/2/3/4 Antibody in Immunocytochemistry (ICC/IF)
          UBE2D1/2/3/4 Monoclonal Antibody (1C7D12), CoraLite® Plus 488
          Product Details
          CL488-68138
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1C7D12
          Immunogen
          UBE2D1/2/3/4 fusion protein Ag2278
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MALKRINKE LSDLARDPPA QCSAGPVGDD MFHWQATIMG PNDSPYQGGV FFLTIHFPTD YPFKPPKVAF TTRIYHPNIN SNGSICLDIL RSQWSPALTI SKVLLSICSL LCDPNPDDPL VPEIARIYKT DRDKYNRISR EWTQKYAM
          Target Information
          Ubiquitination is a post-translational modification pathway or is part of the specific protein-degradation pathway by the 26S proteasome. Ubiquitination of a target protein involves multistep enzymatic reaction catalyzed by a cascade of enzymes including Ub-activating enzymes (E1s), Ub-conjugating enzymes (E2s) and Ub ligases (E3s) (PMID: 23542885). UBE2D family, which has 4 members named as UBE2D1/2/3/4, is an E2 ubiquitin-conjugating enzyme family in the ubiquitin-proteasome system. This antibody can recognize all the four members of UBE2D due to the high homology.
          Bioinformatics
          Protein Aliases: (E3-independent) E2 ubiquitin-conjugating enzyme D1; (E3-independent) E2 ubiquitin-conjugating enzyme D2; (E3-independent) E2 ubiquitin-conjugating enzyme D3; E2 ubiquitin-conjugating enzyme D1; E2 ubiquitin-conjugating enzyme D2; E2 ubiquitin-conjugating enzyme D2B; E2 ubiquitin-conjugating enzyme D3; E2 ubiquitin-conjugating enzyme D4; E2(17)KB 1; E2(17)KB 2; E2(17)KB 2B; E2(17)KB 3; HBUCE1; p53-regulated ubiquitin-conjugating enzyme 1; PAPase; Phosphoarginine phosphatase; predicted protein of HQ2116; SFT; similar to UBC4/5 in yeast; Stimulator of Fe transport; ubc2e ubiquitin conjugating enzyme; UBC4/5 homolog; UbcH5; UBE2D1/UBE2D3/UBE2D3/UBE2D4; UBE2D3; Ubiquitin carrier protein D1; Ubiquitin carrier protein D2; Ubiquitin carrier protein D2B; Ubiquitin carrier protein D3; Ubiquitin carrier protein D4; ubiquitin conjugating enzyme E2 D4 (putative); ubiquitin conjugating enzyme E2D 1; ubiquitin conjugating enzyme E2D 2; ubiquitin conjugating enzyme E2D 3; ubiquitin conjugating enzyme E2D 4 (putative); Ubiquitin-conjugating enzyme E2 D1; Ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2 D2B; Ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2 D4; Ubiquitin-conjugating enzyme E2(17)KB 1; Ubiquitin-conjugating enzyme E2(17)KB 2; Ubiquitin-conjugating enzyme E2(17)KB 2B; Ubiquitin-conjugating enzyme E2(17)KB 3; Ubiquitin-conjugating enzyme E2-17 kDa 1; Ubiquitin-conjugating enzyme E2-17 kDa 2; Ubiquitin-conjugating enzyme E2-17 kDa 2B; Ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 1, UBC4/5 homolog; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme HBUCE1; Ubiquitin-protein ligase D1; Ubiquitin-protein ligase D2; Ubiquitin-protein ligase D2B; Ubiquitin-protein ligase D3; Ubiquitin-protein ligase D4; unnamed protein product
          View more View less
          Gene Aliases: 1100001F19Rik; 9430029A22Rik; AA414951; E2(17)KB1; E2(17)KB2; E2(17)KB3; E217kB; HBUCE1; PUBC1; RGD69425; SFT; Ubc2e; UBC4; UBC4/5; UBC5A; UBC5B; UBC5C; UBCH4; UBCH5; UBCH5A; UBCH5B; UBCH5C; UBCH5D; UBE2D1; UBE2D2; Ube2d2b; UBE2D3; UBE2D4
          View more View less
          UniProt ID: (Human) P51668, (Human) P62837, (Human) P61077, (Human) Q9Y2X8, (Mouse) P61080, (Rat) D3ZDK2, (Rat) P62839, (Mouse) P61079, (Rat) P61078, (Rat) P70711
          View more View less
          Entrez Gene ID: (Human) 7321, (Human) 7322, (Human) 7323, (Human) 51619, (Mouse) 216080, (Rat) 361831, (Rat) 641452, (Mouse) 66105, (Rat) 81920, (Rat) 79435
          View more View less
          Function(s)
          nucleotide binding ubiquitin-protein transferase activity protein binding ATP binding transferase activity ubiquitin protein ligase activity ubiquitin conjugating enzyme activity ligase activity molecular_function ubiquitin-protein ligase
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter protein polyubiquitination ubiquitin-dependent protein catabolic process protein ubiquitination negative regulation of BMP signaling pathway positive regulation of protein ubiquitination cellular response to nutrient levels TORC1 signaling proteasome-mediated ubiquitin-dependent protein catabolic process protein K48-linked ubiquitination positive regulation of protein polyubiquitination negative regulation of TORC1 signaling protein modification process protein autoubiquitination DNA repair protein monoubiquitination apoptotic process cellular response to DNA damage stimulus protein K11-linked ubiquitination protein K6-linked ubiquitination positive regulation of protein targeting to mitochondrion protein K29-linked ubiquitination protein K27-linked ubiquitination protein K63-linked ubiquitination metabolic process biological_process
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline