          • Primary Antibodies ›
          • NOP58 Antibodies
          Proteintech
          NOP58 Recombinant Rabbit Monoclonal Antibody (230058C7), CoraLite® Plus 488
          View all (11) NOP58 antibodies
          Tech Support
          Cite NOP58 Recombinant Rabbit Monoclonal Antibody (230058C7), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          NOP58 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NOP58 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          NOP58 Antibody (CL488-82879) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using CoraLite® Plus 488 NOP58 antibody (CL488-82879, Clone: 230058C7) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NOP58 Antibody in Immunocytochemistry (ICC/IF)
          NOP58 Antibody in Flow Cytometry (Flow)
          NOP58 Recombinant Rabbit Monoclonal Antibody (230058C7), CoraLite® Plus 488
          Product Details
          CL488-82879
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230058C7
          Immunogen
          NOP58 fusion protein Ag5740
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MMAIAPNVTV MVGELVGARL IAHAGSLLNL AKHAASTVQI LGAEKALFRA LKSRRDTPKY GLIYHASLVG QTSPKHKGKI SRMLAAKTVL AIRYDAFGED SSSAMGVENR AKLEARLRTL EDRGIRKISG TGKALAKTEK YEHKSEVKTY DPSGDSTLPT CSKKRKIEQV DKEDEITEKK AKKAKIKVKV EEEEEEKVAE EEETSVKKKK KRGKKKHIKE EPLSEEEPCT STAIASPEKK KKKKKKRENE D
          Target Information
          Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.
          Bioinformatics
          Protein Aliases: NOP58 ribonucleoprotein homolog; Nucleolar protein 5; Nucleolar protein 58; nucleolar protein NOP5/NOP58; unnamed protein product
          View more View less
          Gene Aliases: HSPC120; NOL5; NOP5; NOP5/NOP58; NOP58
          View more View less
          UniProt ID: (Human) Q9Y2X3
          View more View less
          Entrez Gene ID: (Human) 51602
          View more View less
          Function(s)
          TFIID-class transcription factor binding RNA binding protein binding snoRNA binding ATPase binding
          Process(es)
          rRNA processing ribosome biogenesis ribosomal small subunit biogenesis snoRNA localization
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline