Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene is a neural-specific RNA-binding protein that is known to bind to several 3' UTRs, including its own and also that of FOS and ID. The encoded protein may recognize a GAAA motif in the RNA. Three transcript variants encoding two different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV like 4; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; ELAVL 2; ELAVL 4; embryonic lethal, abnormal vision-like 2; Hu antigen B; Hu antigen D; HU-antigen B; HuB; HUD; Nervous system-specific RNA-binding protein Hel-N1; Nervous system-specific RNA-binding protein Mel-N1; RNA binding protein HuB
Gene Aliases: Elav2; ELAVL2; HEL-N1; HELN1; HUB; mel-N1
UniProt ID: (Human) Q12926, (Mouse) Q60899
Entrez Gene ID: (Human) 1993, (Pig) 100153303, (Mouse) 15569, (Rat) 286973
Molecular Function: translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support