Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MPEPLAPESY PESVCEEDVT LALKELDERC EEEEADFSGL SSQDEEEQDG
FRYL gene ontology annotations related to this gene include cell morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALL1-fused gene from chromosome 4p12 protein; DKFZp686E205; FRY like; FRY-like; furry homolog-like; furry-like; mor2 cell polarity protein homolog; Protein furry homolog-like; unnamed protein product
Gene Aliases: 2010313D22Rik; 2310004H21Rik; 2510002A14Rik; 9030227G01Rik; AF4P12; FRYL; KIAA0826; mKIAA0826; MOR2; PCBS
UniProt ID: (Human) O94915
Entrez Gene ID: (Human) 285527, (Mouse) 72313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support