Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LMGLYHGQV LCKKNKFGAP QKDVKHLAIL GAGLMGAGIA QVSVDKGLKT ILKDATLTAL DRGQQQVFKG LNDKVKKKAL TSFERDSIFS NLTGQLDYQG FEKADMVIEA VFEDLSLKHR VLKEVEAVIP DHCIFASNTS ALPISEIAAV SKRPEKVIGM HYFSPVDKMQ LLEIITTEKT SKDTSASAVA VGLKQGKVII VVKDGPGFYT TRCLAPMMSE VIRILQEGVD PKKLDSLTTS FGFPVGAATL VDEVGVDVAK HVAEDLGKVF GERFGGGNPE LLTQMVSKGF LGRKSGKGFY IYQEGVKRKD LNSDMDSILA SLKLPPKSEV SSDEDIQFRL VTRFVNEAVM CLQEGILATP AEGDIGAVFG LGFPPCLGGP FRFVDLYGAQ KIVDRLKKYE AAYGKQFTPC QLLADHANSP NKKFYQ (339-763 aa encoded by BC009235)
This gene encodes the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities. Mutations in this gene result in trifunctional protein deficiency or LCHAD deficiency. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-ketoacyl-Coenzyme A (CoA) thiolase, alpha subunit; 3-oxoacyl-CoA thiolase; 78 kDa gastrin-binding protein; gastrin-binding protein; hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit; hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit; long-chain 2-enoyl-CoA hydratase; long-chain-3-hydroxyacyl-CoA dehydrogenase; mitochondrial long-chain 2-enoyl-Coenzyme A (CoA) hydratase, alpha subunit; mitochondrial long-chain L-3-hydroxyacyl-Coenzyme A (CoA) dehydrogenase, alpha subunit; mitochondrial trifunctional enzyme, alpha subunit; mitochondrial trifunctional protein, alpha subunit; monolysocardiolipin acyltransferase; TP-alpha; unnamed protein product
Gene Aliases: ECHA; GBP; LCEH; LCHAD; MLCL AT; MTPA; RGD1560655; TP-ALPHA
UniProt ID: (Human) P40939, (Mouse) Q8BMS1, (Rat) Q64428
Entrez Gene ID: (Human) 3030, (Mouse) 97212, (Rat) 170670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support