Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RSQRLSTAS VIQARYQGLV DGRKGAHAPQ ELPQAKGSDD ELFLGGSNQD SVQGLGRAVC PAGGEAGLGG CSSSTNGSSG GCEQELGLDL SKKSPPLPPA TPGPHLTPDD AAQLSDSQHG SPPAASAPPV ANSASYSELG GTPDEPMDLE GAEDNHLSLL EAPGGQPRKS LRHSTRKKEW GKKEPVAGSP FERREAGPKG PCPGEEGEGV GDRVPNGILA SGAGPSGPYG EPPYPCKEEE ENGKDASEDS AQSGSEGGSG HASAHYMYRQ E (161-430 aa encoded by BC094787)
HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Em:AP000557.1; Hic-2; Hic-3; HIC1-related gene on chromosome 22 protein; Hypermethylated in cancer 2 protein; Zinc finger and BTB domain-containing protein 30
Gene Aliases: HIC2; HRG22; KIAA1020; ZBTB30; ZNF907
UniProt ID: (Human) Q96JB3
Entrez Gene ID: (Human) 23119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support