    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ly-6G/Ly-6C Antibodies
          Invitrogen
          Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5)
          44 References
          View all (65) Ly-6G/Ly-6C antibodies
          Tech Support
          Cite Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5)
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Ly-6G/Ly-6C Antibody (MA1-10401) in Flow
          Separation of murine Ly6G positive cells (red-filled) from Ly6G negative cells (black-dashed) in flow cytometry analysis (surface staining) of murine splenocyte suspension stained using anti-mouse Ly6G (RB6-8C5) purified Monoclonal antibody (Product # MA1-10401) (low endotoxin, concentration in sample 9 µg/mL) DAR PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5)
          Product Details
          MA1-10401
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1 µg/mL
          -
          Immunohistochemistry (IHC)
          -
          View 2 publications 2 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-Dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-Dependent
          View 3 publications 3 publications
          Flow Cytometry (Flow)
          1-4 µg/mL
          View 29 publications 29 publications
          Immunoprecipitation (IP)
          1-2 µg/100-500 µg protein
          -
          Functional Assay (Functional)
          Assay-dependent
          View 1 publication 1 publication
          Miscellaneous PubMed (Misc)
          -
          View 9 publications 9 publications
          Product Specifications
          Species Reactivity
          Mouse
          Published species
          Mouse
          Host/Isotype
          Rat / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          RB6-8C5
          Immunogen
          Murine granulocytes
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FITC
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4
          Contains
          no preservative
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_11152791
          Product Specific Information
          This antibody detects Ly6G component of Gr-1 antigen, a commonly used surface marker of neutrophils.
          Functional Application: Depletion.
          Target Information
          Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
          Bioinformatics
          Protein Aliases: Granulocyte Marker; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN; unnamed protein product
          View more View less
          Gene Aliases: Gr-1; Gr1; Ly-6C; Ly-6C.2; Ly-6C1; Ly-6C2; Ly-6G; Ly6c
          View more View less
          UniProt ID: (Mouse) P0CW02, (Mouse) P0CW03, (Mouse) P35461
          View more View less
          Entrez Gene ID: (Mouse) 17067, (Mouse) 100041546, (Mouse) 546644
          View more View less
          Function(s)
          acetylcholine receptor inhibitor activity acetylcholine receptor binding
          Process(es)
          acetylcholine receptor signaling pathway
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-65f6797749-ftzwf:80/100.66.76.64:80.
          git-commit: 74bf1af917bc7c7f736041a944cb80ce7b99a823
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.0-PolychromeNPI-91