          • Primary Antibodies ›
          • N-sulfated Heparan Sulfate Antibodies
          Invitrogen
          N-sulfated Heparan Sulfate Monoclonal Antibody (HepSS-1), PE
          View all (3) N-sulfated Heparan Sulfate antibodies
          Tech Support
          Cite N-sulfated Heparan Sulfate Monoclonal Antibody (HepSS-1), PE
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          N-sulfated Heparan Sulfate Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          N-sulfated Heparan Sulfate Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          N-sulfated Heparan Sulfate Antibody (MA574408) in Flow
          Flow cytometry analysis of N-sulfated Heparan Sulfate in EA.hy926 cells (red-filled). Samples were incubated with monoclonal N-sulfated Heparan Sulfate (Product # MA574408) using a dilution of 10 μg/mL, followed by PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          N-sulfated Heparan Sulfate Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA574408
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          2-5 µg/mL
          -
          Product Specifications
          Species Reactivity
          Chemical
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          HepSS-1
          Immunogen
          MethA murine fibrosarcoma.
          Conjugate
          PE PE PE
          • Unconjugated
          • Biotin 
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Heparan sulfate (HS) is a glycosaminoglycan that is ubiquitously expressed on cell surfaces and in the extracellular matrix and basement membrane. Each HS molecule is a linear polysaccharide composed of repeating disaccharides of hexuronic acid and d-glucosamine that can exhibit immense structural diversity due to substitution to varying extents with sulfate groups and epimerization of glucuronic acid to iduronic acid, with areas of high sulfation and glucuronic acid epimerization being co-located in hot spots throughout the molecule. HS is structurally related to heparin, an extremely highly sulfated form of HS that is restricted to mast cells. The biosynthesis and modification of HS chains is thought to take place within the endoplasmic reticulum, Golgi apparatus, and trans Golgi network, which in the end produce unique HS chains that are covalently attached to a range of core proteins to form HS-proteoglycans [PMID: 24391644].
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: Heparan sulfate
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0