          • Primary Antibodies ›
          • SNX13 Antibodies
          Invitrogen
          SNX13 Polyclonal Antibody
          View all (3) SNX13 antibodies
          Tech Support
          Cite SNX13 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          SNX13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          SNX13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          SNX13 Antibody (PA5-121589) in IHC (P)
          Immunohistochemistry analysis of SNX13 in paraffin-embedded Human Pancreas tissue. Samples were incubated with polyclonal antibody (Product # PA5-119427) at a dilution of 30 µg/mL, followed by HRP-Linked Caprine Anti-Rabbit IgG using a dilution of 2 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SNX13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SNX13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SNX13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SNX13 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SNX13 Antibody in Western Blot (WB)
          SNX13 Antibody in Western Blot (WB)
          SNX13 Polyclonal Antibody
          Product Details
          PA5-121589
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-3 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-30 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein Sorting Nexin 13. The antigen corresponds to amino acid range 97-284 of the target protein.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915161
          Target Information
          May be involved in several stages of intracellular trafficking. May play a role in endosome homeostasis. Acts as a GAP for Galphas.
          Bioinformatics
          Protein Aliases: RGS domain- and PHOX domain-containing protein; RGS-PX1; Sorting nexin-13; unnamed protein product
          View more View less
          Gene Aliases: KIAA0713; RGS-PX1; SNX13
          View more View less
          UniProt ID: (Human) Q9Y5W8
          View more View less
          Entrez Gene ID: (Human) 23161
          View more View less
          Function(s)
          lipid binding phosphatidylinositol-3-phosphate binding phosphatidylinositol binding
          Process(es)
          intracellular protein transport negative regulation of signal transduction protein transport positive regulation of GTPase activity
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline