          • Primary Antibodies ›
          • Gtf2h5 Antibodies
          Invitrogen
          Gtf2h5 Polyclonal Antibody
          View all (3) Gtf2h5 antibodies
          Tech Support
          Cite Gtf2h5 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          Gtf2h5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Gtf2h5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Gtf2h5 Antibody (PA5-121936) in ICC/IF
          Immunocytochemistry analysis of Gtf2h5 in HepG2 cells using Gtf2h5 Polyclonal Antibody (Product # PA5-121935) at a dilution of 20 µg/mL, followed by secondary antibody staining with 2 µg/mL AF488-Linked Caprine Anti-Rabbit IgG Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Gtf2h5 Antibody in Immunocytochemistry (ICC/IF)
          Gtf2h5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gtf2h5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gtf2h5 Antibody in Western Blot (WB)
          Gtf2h5 Polyclonal Antibody
          Product Details
          PA5-121936
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          20 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Met1-Lys71 (Accession Q6ZYL4), with N-terminal His Tag
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915508
          Target Information
          Component of the general transcription and DNA repair factor IIH (TFIIH) core complex, which is involved in general and transcription-coupled nucleotide excision repair (NER) of damaged DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II. In NER, TFIIH acts by opening DNA around the lesion to allow the excision of the damaged oligonucleotide and its replacement by a new DNA fragment. In transcription, TFIIH has an essential role in transcription initiation. When the pre-initiation complex (PIC) has been established, TFIIH is required for promoter opening and promoter escape. Phosphorylation of the C-terminal tail (CTD) of the largest subunit of RNA polymerase II by the kinase module CAK controls the initiation of transcription. Necessary for the stability of the TFIIH complex and for the presence of normal levels of TFIIH in the cell.
          Bioinformatics
          Protein Aliases: General transcription factor IIH polypeptide 5; General transcription factor IIH subunit 5; general transcription factor IIH, polypeptide 5; TFB5 ortholog; TFIIH basal transcription factor complex TTD-A subunit; TFIIH basal transcription factor complex TTDA subunit; TFIIH subunit p8
          View more View less
          Gene Aliases: bA120J8.2; C6orf175; GTF2H5; TFB5; TFIIH; TGF2H5; TTD; TTD-A; TTD3; TTDA
          View more View less
          UniProt ID: (Human) Q6ZYL4
          View more View less
          Entrez Gene ID: (Human) 404672
          View more View less
          Function(s)
          protein binding
          Process(es)
          maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA) DNA repair nucleotide-excision repair nucleotide-excision repair, preincision complex assembly transcription elongation from RNA polymerase I promoter transcription from RNA polymerase II promoter transcription initiation from RNA polymerase II promoter cellular response to DNA damage stimulus cellular response to gamma radiation
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline