          • Primary Antibodies ›
          • A26C2 Antibodies
          Invitrogen
          A26C2 Polyclonal Antibody
          View all (14) A26C2 antibodies
          Tech Support
          Cite A26C2 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          A26C2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          A26C2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          A26C2 Antibody (PA5-122306) in WB
          Western blot analysis of A26C2 in recombinant protein. Samples were incubated in polyclonal A26C2 antibody (Product # PA5-122305). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          A26C2 Antibody in Western Blot (WB)
          A26C2 Polyclonal Antibody
          Product Details
          PA5-122306
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.5-5 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-20 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          5-20 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant POTE Ankyrin Domain Family, Member G (POTEG), Leu177-Asp330 (Accession Q6S5H5).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915878
          Target Information
          Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
          Bioinformatics
          Protein Aliases: ANKRD26-like family C member 2; cancer/testis antigen family 104, member 4; POTE ankyrin domain family member G; POTE-14; Prostate, ovary, testis-expressed protein on chromosome 14; protein expressed in prostate, ovary, testis, and placenta 14
          View more View less
          Gene Aliases: A26C2; ACTBL1; CT104.4; POTE-14; POTE14; POTE14alpha; POTE22; POTEG
          View more View less
          UniProt ID: (Human) Q6S5H5
          View more View less
          Entrez Gene ID: (Human) 404785
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline