Search Thermo Fisher Scientific
Recommended positive controls: Jurkat, Raji, K562.
Predicted reactivity: Rhesus Monkey (98%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Kruppel-like zinc finger proteins such as ZNF189 contain a conserved stretch of 7 amino acids that connects a variable number of DNA-binding zinc finger repeats of the cys(2)his(2) (C2H2) type (summarized by Odeberg et al., 1998 [PubMed 9653648]). Approximately 30% of human Kruppel-like zinc finger proteins contain an N-terminal Kruppel-associated box (KRAB) domain. The KRAB domain consists of approximately 75 amino acids that may be subdivided into an A box, which is present in every KRAB domain and is essential for transcriptional repression, and a B box, which is not always present.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: Zinc finger protein 189
Gene Aliases: ZNF189
UniProt ID: (Human) O75820
Entrez Gene ID: (Human) 7743
Molecular Function: DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator