Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MATAPYNYS YIFKYIIIGD MGVGKSCLLH QFTEKKFMAD CPHTIGVEFG TRIIEVSGQK IKLQIWDTAG QERFRAVTRS YYRGAAGALM VYDITRRSTY NHLSSWLTDA RNLTNPNTVI ILIGNKADLE AQRDVTYEEA KQFAEENGLL FLEASAKTGE NVEDAFLEAA KKIYQNIQDG SLDLNAAESG VQHKPSAPQG GRLTSEPQPQ REGCGC (1-215 aa encoded by B C006081)
Localized to biosynthetic and endosomal compartments, Rab14 is involved in the biosynthetic/recycling pathway between the Golgi and endosomal compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA165P4.3 (member RAS oncogene family); F protein-binding protein 1; GTPase Rab14; Rab protein family member; RAB-14; RAB14; small GTP binding protein RAB14; unnamed protein product; viral protein-binding protein; FBP1
Gene Aliases: 0610030G24Rik; 2810475J17Rik; A830021G03Rik; D030017L14Rik; FBP; RAB-14
UniProt ID: (Human) P61106, (Mouse) Q91V41, (Rat) P61107
Entrez Gene ID: (Human) 51552, (Mouse) 68365, (Rat) 94197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support