Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • STX12 Antibodies

          Proteintech

          Syntaxin 12 Polyclonal Antibody

          4 References
          View all (12) STX12 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Syntaxin 12 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (23)
          Syntaxin 12 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Syntaxin 12 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Syntaxin 12 Antibody (14259-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using STX12 antibody 14259-1-AP at 1:50 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Syntaxin 12 Antibody in Immunocytochemistry (ICC/IF)
          Syntaxin 12 Antibody in Immunocytochemistry (ICC/IF)
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Western Blot (WB)
          Syntaxin 12 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Syntaxin 12 Polyclonal Antibody

          Product Details

          14259-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Syntaxin 12 Fusion Protein Ag5532 (1-254 aa encoded by BC046999)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          25.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody recognizes a major 39 kDa band of endogenous STX12. In some cases, two additional bands of 33 kDa and 66 kDa could also be detected, which most likely represent a degradation product and a SNARE complex respectively.

          Immunogen sequence: MSYGPLDMY RNPGPSGPQL RDFSSIIQTC SGNIQRISQA TAQIKNLMSQ LGTKQDSSKL QENLQQLQHS TNQLAKETNE LLKELGSLPL PLSTSEQRQQ RLQKERLMND FSAALNNFQA VQRRVSEKEK ESIARARAGS RLSAEERQRE EQLVSFDSHE EWNQMQSQED EVAITEQDLE LIKERETAIR QLEADILDVN QIFKDLAMMI HDQGDLIDSI EANVESSEVH VERATEQLQR AAYYQKKSRK KMCIL (1-254 aa encoded by BC046999)

          Target Information

          SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MGC51957; STX12; syntaxin 13; syntaxin-13; unnamed protein product

          View more View less

          Gene Aliases: STX13; STX14; Syn13

          View more View less

          UniProt ID: (Human) Q86Y82, (Mouse) Q9ER00, (Rat) G3V7P1

          View more View less

          Entrez Gene ID: (Human) 23673, (Mouse) 100226, (Rat) 65033

          View more View less

          Function(s)
          SNARE binding SNAP receptor activity protein binding
          Process(es)
          autophagosome assembly intracellular protein transport vesicle fusion vesicle-mediated transport endocytic recycling cholesterol efflux protein stabilization membrane fusion vesicle organization synaptic vesicle to endosome fusion organelle fusion regulation of postsynaptic membrane neurotransmitter receptor levels
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-5d76f9f486-2dllf:80/100.66.78.26:80.
          git-commit: 08b979eaab39376f31a39358ffa32de998e3c7f7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.6.0-offline-prod