          • Primary Antibodies ›
          • MRAS Antibodies
          OriGene
          MRAS Monoclonal Antibody (OTI3C4), TrueMAB™
          View all (14) MRAS antibodies
          Tech Support
          Cite MRAS Monoclonal Antibody (OTI3C4), TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          MRAS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MRAS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          MRAS Antibody (TA502864S) in ICC/IF
          Anti-MRAS mouse monoclonal antibody (TA502864) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY MRAS(RC212259). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MRAS Antibody in Immunocytochemistry (ICC/IF)
          MRAS Antibody in Western Blot (WB)
          MRAS Antibody in Flow Cytometry (Flow)
          MRAS Monoclonal Antibody (OTI3C4), TrueMAB™
          Product Details
          TA502864S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI3C4
          Immunogen
          Full length human recombinant protein of human MRAS produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.69 mg/mL
          Amount
          20.7 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 1% BSA, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
          Bioinformatics
          Protein Aliases: GTP-binding protein; MRAS; muscle and microspikes RAS; Ras-related protein M-Ras; Ras-related protein R-Ras3; small GTP binding protein; unnamed protein product
          View more View less
          Gene Aliases: M-RAs; MRAS; NS11; R-RAS3; RRAS3
          View more View less
          UniProt ID: (Human) O14807
          View more View less
          Entrez Gene ID: (Human) 22808
          View more View less
          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding hydrolase activity GDP binding GTP-dependent protein binding
          Process(es)
          signal transduction Ras protein signal transduction actin cytoskeleton organization cellular response to leukemia inhibitory factor
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline