Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MAAAGAPDGM EEPGMDTEAE TVATEAPARP VNCLEAEAAA GAAAEDSGAA RGSLQPAPAQ PPGDPAAQAS VSNGEDAGGG AGRELVDLKI IWNKTKHDVK FPLDSTGSEL KQKIHSITGL PPAMQKVMYK GLVPEDKTLR EIKVTSGAKI MVVGSTINDV LAVNTPKDAA QQDAKAEENK KEPLCRQKQH RKVLDKGKPE DVMPSVKGAQ ERLPTVPLSG MYNKSGGKVR LTFKLEQDQL WIGTKERTEK LPMGSIKNVV SEPIEGHEDY
May play a role as NF-kappa-B regulator. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ubiquitin domain-containing protein UBFD1; Ubiquitin-binding protein homolog; unnamed protein product
Gene Aliases: AI467302; D7Wsu105e; D7Wsu128e; UBFD1; UBPH
UniProt ID: (Human) O14562, (Mouse) Q78JW9
Entrez Gene ID: (Human) 56061, (Mouse) 28018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support