Technical Resources - Media Formulations
21083 - Leibovitz's L-15, no phenol red
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.0200.02.6666667
L-Alanine89.0225.02.52809
L-Arginine174.0500.02.8735633
L-Asparagine132.0250.01.8939394
L-Cysteine121.0120.00.9917355
L-Glutamine146.0300.02.0547945
L-Histidine155.0250.01.6129032
L-Isoleucine131.0250.01.908397
L-Leucine131.0125.00.9541985
L-Lysine146.075.00.51369864
L-Methionine149.075.00.5033557
L-Phenylalanine165.0125.00.75757575
L-Serine105.0200.01.9047619
L-Threonine119.0300.02.5210085
L-Tryptophan204.020.00.09803922
L-Tyrosine181.0300.01.6574585
L-Valine117.0100.00.85470086
Vitamins
Choline chloride140.01.00.007142857
D-Calcium pantothenate477.01.00.002096436
Folic Acid441.01.00.0022675737
Niacinamide122.01.00.008196721
Pyridoxine hydrochloride206.01.00.004854369
Riboflavin 5'-phosphate Na478.00.12.0920503E-4
Thiamine monophosphate442.01.00.0022624435
i-Inositol180.02.00.011111111
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0140.01.2612612
Magnesium Chloride (anhydrous)95.093.70.9863158
Magnesium Sulfate (MgSO4) (anhyd.)120.097.670.8139166
Potassium Chloride (KCl)75.0400.05.3333335
Potassium Phosphate monobasic (KH2PO4)136.060.00.44117647
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.0190.01.3380282
Other Components
D+ Galactose180.0900.05.0
Sodium Pyruvate110.0550.05.0
参照
Related Products: