+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Phadia Laboratory Systems
Your Laboratory Depends on Accurate and Reproducible Results
PhadiaTM Laboratory Systems are designed to help your laboratory deliver successful test results, quickly and efficiently. With high sensitivity and specificity, these highly automated systems use fluorescence enzyme immunoassay (FEIA) technology to offer the superior ability to detect and quantitatively determine specific antibodies in blood.
Matching Lab Goals
By processing samples from test requests to final results with minimal interventions, Phadia Laboratory Systems reduce hands-on time and improve your workflow:
  • Easier audits and set-up with barcode identification of all reagents
  • User efficiency and minimized errors with mainframe worklist downloads, barcode sample identification, and on-board dilution
  • Faster loading times with on-board storage of all reagents
  • Ability to run ImmunoCAPTM and EliATM assays on the same system*
*The Phadia 1000 instrument processes ImmunoCAP assay exclusively
Intuitive Data Management Software
Designed to simplify and optimize daily operation
Phadia Prime software optimizes processing of ImmunoCAP and EliA assays on Phadia Laboratory Systems to aid in the diagnosis of allergic and autoimmune diseases. Innovative features of Phadia Prime that help your laboratory meet today’s workflow demands include:
time
Intuitive, real-time management of reagent inventory
specsens
Extended audit trail
report
graph
Enterprise Resource Planning (ERP) integration
Cog
Two-way LIS channel communication and dual LIS channel capabilities
tube
Test prioritization by adult/pediatric samples
2 Test tube
Integrated Quantity Not Sufficient (QNS) handling
paper icon
Customizable reflex testing capabilities to support diagnostic pathways
This state-of-the-art data management software allows efficient management of multiple Phadia instruments. Phadia Prime software, an integral part of Phadia Laboratory Systems, continuously helps improve productivity in clinical laboratories.