            • カスタム製品とプロジェクト
          • ELISA Kits ›
          • Luteinizing Hormone ELISA Kits
          Invitrogen
          Human Luteinizing Hormone ELISA Kit
          2 References
          View all (5) Luteinizing Hormone ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          Human Luteinizing Hormone ELISA Kit
          Group 53 Created with Sketch.
          Human Luteinizing Hormone ELISA Kit
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human Luteinizing Hormone ELISA Kit
          ELISA Standard curve of Luteinizing Hormone using Human Luteinizing Hormone ELISA Kit (Product # EHLH). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human Luteinizing Hormone ELISA Kit
          Human Luteinizing Hormone ELISA Kit (EHLHX10)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          140 pg/mL
          Assay range
          140-10,000 pg/mL
          Sample type/volume
          Plasma
          50 µL
          Serum
          50 µL
          Supernatant
          100 µL
          Hands-on time
          1 hr 20 min
          Time-to-result
          4 hr 45 min
          Homogenous (no wash)
          No
          Interassay CV
          <12%
          Intraassay CV
          <10%
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          Pre-coated 96 well plate
          Standard
          Assay Diluent concentrate
          Biotinylated Detection Antibody
          SAV-HRP
          Wash Buffer
          Chromogen
          Stop Solution
          Adhesive Plate Covers
          Shipping conditions
          Wet ice
          Storage
          Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
          Protein name
          Luteinizing Hormone
          Protein family
          Other Proteins
          Species (tested)
          Human
          Assay kit format
          Sandwich ELISA Kit
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          The Human Luteinizing Hormone (Hu LH) ELISA quantitates Hu LH in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu LH.
          Principle of the method
          The Human LH solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
          Rigorous validation
          Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
          Target information
          LH is a member of the glycoprotein hormone family, Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
          Bioinformatics
          Gene aliases : CG-ALPHA, CGA, CGB4, FSHA, GPA1, GPHa, GPHA1, HCG, HH23, LHA, LHB, LSH-B, LSH-beta, TSHA
          Gene ID : (Human) 1081, (Human) 3972
          Gene symbol : CGA, LHB
          Protein Aliases : Anterior pituitary glycoprotein hormones common subunit alpha, CG-alpha, CGA, Choriogonadotropin alpha chain, Chorionic gonadotrophin subunit alpha, chorionic gonadotropin, alpha polypeptide, Follicle-stimulating hormone alpha chain, follicle-stimulating hormone alpha subunit, Follitropin alpha chain, FSH-alpha, Glycoprotein hormones alpha chain, interstitial cell stimulating hormone, beta chain, LH-B, LSH-alpha, Luteinizing hormone alpha chain, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, Luteinizing hormone subunit beta, Lutropin alpha chain, Lutropin beta chain, LSH-B, LSH-beta, follitropin, FSH-B, FSH-beta, hLHB, Lutropin subunit beta, Thyroid-stimulating hormone alpha chain, Thyrotropin alpha chain, TSH-alpha, unnamed protein product
          UniProt ID (Human) P01215, (Human) P01229
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline