Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Human CD79b ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human CD79B.
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : AGM6, B29, IGB, Igbeta
Gene ID : (Human) 974
Gene symbol : CD79B
Protein Aliases : Ag receptor complex, B-cell-specific glycoprotein B29, CD79, Ig beta, RP23-418O11.7, CD79b antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, extracellular domain, Ig-beta, immunoglobulin-associated B29 protein, unnamed protein product
UniProt ID (Human) P40259
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support