Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Human Luteinizing Hormone Rapid ELISA Kit quantitates Luteinizing Hormone in serum and plasma
LH is a member of the glycoprotein hormone family, Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CG-ALPHA, CGA, CGB4, FSHA, GPA1, GPHa, GPHA1, HCG, HH23, LHA, LHB, LSH-B, LSH-beta, TSHA
Gene ID : (Human) 1081, (Human) 3972
Gene symbol : CGA, LHB
Protein Aliases : Anterior pituitary glycoprotein hormones common subunit alpha, CG-alpha, CGA, Choriogonadotropin alpha chain, Chorionic gonadotrophin subunit alpha, chorionic gonadotropin, alpha polypeptide, Follicle-stimulating hormone alpha chain, follicle-stimulating hormone alpha subunit, Follitropin alpha chain, follitropin, FSH-B, FSH-beta, hLHB, FSH-alpha, Glycoprotein hormones alpha chain, interstitial cell stimulating hormone, beta chain, LH-B, LSH-alpha, Luteinizing hormone alpha chain, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, Luteinizing hormone subunit beta, Lutropin alpha chain, Lutropin beta chain, Lutropin subunit beta, Thyroid-stimulating hormone alpha chain, Thyrotropin alpha chain, TSH-alpha, unnamed protein product
UniProt ID (Human) P01215, (Human) P01229
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support