Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Mouse Procollagen I C-Peptide ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of mouse PCPE-1 in cell culture supernatants, plasma, and serum samples.
Collagen type I is present in soft connective tissue and bone where it represents approximately 90% of the organix matrix. Collagen type I is produced from procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. The propeptide levels can be measured in serum as indicators of collagen production. Elevated values of these propeptides in serum are found in patients with various bone metabolism defects. Procollagen type I C-terminal propeptide (PICP) has a molecular weight of approximately 100 kDa and the normal range in serum depends on age and sex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Col1a-1, Cola-1, Cola1, Mov-13, Mov13
Gene ID : (Mouse) 12842
Gene symbol : Col1a1
Protein Aliases : alpha-1 type 1 collagen, alpha-1 type I collagen, C-terminal Collagen, PICP, Procol, Procollagen, pro-alpha-1 type I collagen, procollagen, type I, alpha 1
UniProt ID (Mouse) P11087
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support