Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Rat Proprotein Convertase 9/PCSK9 Rapid ELISA Kit quantitates PCSK9 in serum and plasma
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : NARC-1, Narc1, PC9, Pcsk9
Gene ID : (Rat) 298296
Gene symbol : Pcsk9
Protein Aliases : hypercholesterolemia, OTTHUMP00000009833, NARC-1, neural apoptosis regulated convertase 1, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase PC9, Proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
UniProt ID (Rat) P59996
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support