Search
Nalgene polypropylene bottles are something special. Our bottles are made from a polypropylene copolymer material which improves low temperature performance. From -40°C to the autoclave, these bottles are suitable for a wide range of laboratory applications. Polypropylene copolymer bottles provide very good chemical compatibility with a wide variety of lab reagents including most acids, bases, alcohols, aldehydes, esters, and aliphatic hydrocarbons.
For chemical compatibility ratings by specific chemical and application temperature, consult the Nalgene Chemical Resistance Guide, or request a copy of the printed wall chart. Polypropylene copolymer bottles are made only from high quality laboratory grade resin materials that comply with a number of regulatory specifications including: USP Class VI, EP monographs, EU food-contact directives, CONEG, RoHS, CA Prop 65, SARA Title III Sec. 313, and 21 CFR pt 177.1520 for food contact. For regulatory claims and certifications applicable to specific Nalgene bottles, please contact Nalgene Regulatory Support. Guaranteed leakproof* where noted.
For help selecting the Nalgene bottle that's perfect for your lab applications, use our online selection guide or for personal assistance, contact the Nalgene Technical Support team.
| Narrow-Mouth Round Lab Quality Bottle (2006-series) | Narrow-Mouth Round Economy Bottle (2087-series) | Narrow-Mouth Large Round Bottle (2203-series) | Narrow-Mouth Large Round Amber Bottle (2204-series) |
| Traditional reusable lab bottle for long-term use and critical applications | Lower cost, lighter weight, thinner wall for light-duty applications | Larger size, rugged lab quality construction | Amber color protects light-sensitive solutions, larger size, rugged lab quality construction |
![]() | ![]() | ![]() | ![]() |
| PPCO bottle PP Closure | PPCO bottle white PP Closure | PPCO bottle PP Closure | Amber PPCO bottle amber or white closure |
| Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber |
| 4mL–1L | 30mL–1L | 2L–8L | 2L–8L |
| Leakproof* | Leakproof* | Leakproof* | Leakproof* |
| No graduations | No graduations | No graduations | No graduations |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40oC to 121oC |
| Autoclavable | Autoclavable | Autoclavable | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
| Also available in bulk, unassembled | Also available in bulk, unassembled |
| Narrow-Mouth Round Lab Quality Bottle (2006-series) | Narrow-Mouth Round Economy Bottle (2087-series) | Narrow-Mouth Large Round Bottle (2203-series) | Narrow-Mouth Large Round Amber Bottle (2204-series) | |
| Acids, dilute or weak | E | E | E | E |
| Acids^ strong, concentrated | G | G | G | G |
| Alcohols, aliphatic | E | E | E | E |
| Aldehydes | G | G | G | G |
| Bases/alkali | E | E | E | E |
| Esters | G | G | G | G |
| Hydrocarbons, aliphatic | G | G | G | G |
| Hydrocarbons, aromatic | N | N | N | N |
| Hydrocarbons, halogenated | N | N | N | N |
| Ketones, aromatic | N | N | N | N |
| Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20 oC. |
| G | Little or no damage after 30 days constant exposure at 20oC. |
| F | Some effect after 7 days constant exposure at 20oC. |
| N | Not recommended. Immediate damage may occur. |
| Wide-Mouth Round Lab Quality Bottle (2105-series) | Wide-Mouth Round Economy Bottle (2187-series) | Large-Wide Mouth Round Bottle (2121-series) |
| Traditional reusable lab bottle for long-term use and critical applications | Lower cost, lighter weight, thinner wall for light-duty applications | Larger size, rugged lab quality construction |
![]() | ![]() | ![]() |
| PPCO bottle PP Closure | PPCO bottle PP Closure | PPCO bottle white pp closure |
| Milky white translucent | Milky white translucent | Milky white translucent |
| 30mL–1L | 30mL–1L | 2L–4L |
| Leakproof* | Leakproof* | Not liquid leakproof |
| No graduations | No graduations | No graduations |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40oC to 121oC |
| Autoclavable | Autoclavable | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free |
| Also available in bulk, unassembled | Also available in bulk, unassembled |
| Wide-Mouth Round Lab Quality Bottle (2105-series) | Wide-Mouth Round Economy Bottle (2187-series) | Large Wide-Mouth Round Bottle (2121-series) | |
| Acids, dilute or weak | E | E | E |
| Acids^ strong, concentrated | G | G | G |
| Alcohols, aliphatic | E | E | E |
| Aldehydes | G | G | G |
| Bases/alkali | E | E | E |
| Esters | G | G | G |
| Hydrocarbons, aliphatic | G | G | G |
| Hydrocarbons, aromatic | N | N | N |
| Hydrocarbons, halogenated | N | N | N |
| Ketones, aromatic | N | N | N |
| Oxidizing agents, strong | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20 oC |
| G | Little or no damage after 30 days constant exposure at 20oC |
| F | Some effect after 7 days constant exposure at 20oC |
| N | Not recommended. Immediate damage may occur. |
| Narrow-Mouth Square Lab Quality Bottle (2016-series) | Wide-Mouth Square Lab Quality Bottle (2110-series) | Large Wide-Mouth Square Bottle (2122-series) |
| Space-saving square shape, narrow mouth, graduated | Space-saving square shape, wide mouth | Space-saving square shape, wide mouth, large capacity, molded hand grip |
![]() | ![]() | ![]() |
| PPCO bottle/ PP Closure | PPCO bottle/ PP Closure | PPCO bottle/ white PP closure |
| Milky white translucent | Milky white translucent | Milky white translucent |
| 30mL–1L | 60mL–1L | 4L |
| L* | Leakproof* | Not liquid leakproof |
| Molded graduations | No graduations | No graduations |
| Narrow mouth | Wide mouth | Wide mouth |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40°C to 121°C |
| Autoclavable | Autoclavable | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free |
| Narrow-Mouth Square Lab Quality Bottle (2016-series) | Wide-Mouth Square Lab Quality Bottle (2187-series) | Large Wide-Mouth Square Bottle (2122-series) | |
| Acids, dilute or weak | E | E | E |
| Acids^ strong, concentrated | G | G | G |
| Alcohols, aliphatic | E | E | E |
| Aldehydes | G | G | G |
| Bases/alkali | E | E | E |
| Esters | G | G | G |
| Hydrocarbons, aliphatic | G | G | G |
| Hydrocarbons, aromatic | N | N | N |
| Hydrocarbons, halogenated | N | N | N |
| Ketones, aromatic | N | N | N |
| Oxidizing agents, strong | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20oC. |
| G | Little or no damage after 30 days constant exposure at 20oC. |
| F | Some effect after 7 days constant exposure at 20oC. |
| N | Not recommended. Immediate damage may occur. |
| Narrow-Mouth Round Lab Quality Bottle (2006-series) | Narrow-Mouth Round Economy Bottle (2087-series) | Narrow-Mouth Large Round Bottle (2203-series) | Narrow-Mouth Large Round Amber Bottle (2204-series) |
| Traditional reusable lab bottle for long-term use and critical applications | Lower cost, lighter weight, thinner wall for light-duty applications | Larger size, rugged lab quality construction | Amber color protects light-sensitive solutions, larger size, rugged lab quality construction |
![]() | ![]() | ![]() | ![]() |
| PPCO bottle PP Closure | PPCO bottle white PP Closure | PPCO bottle PP Closure | Amber PPCO bottle amber or white closure |
| Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber |
| 4mL–1L | 30mL–1L | 2L–8L | 2L–8L |
| Leakproof* | Leakproof* | Leakproof* | Leakproof* |
| No graduations | No graduations | No graduations | No graduations |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40oC to 121oC |
| Autoclavable | Autoclavable | Autoclavable | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
| Also available in bulk, unassembled | Also available in bulk, unassembled |
| Narrow-Mouth Round Lab Quality Bottle (2006-series) | Narrow-Mouth Round Economy Bottle (2087-series) | Narrow-Mouth Large Round Bottle (2203-series) | Narrow-Mouth Large Round Amber Bottle (2204-series) | |
| Acids, dilute or weak | E | E | E | E |
| Acids^ strong, concentrated | G | G | G | G |
| Alcohols, aliphatic | E | E | E | E |
| Aldehydes | G | G | G | G |
| Bases/alkali | E | E | E | E |
| Esters | G | G | G | G |
| Hydrocarbons, aliphatic | G | G | G | G |
| Hydrocarbons, aromatic | N | N | N | N |
| Hydrocarbons, halogenated | N | N | N | N |
| Ketones, aromatic | N | N | N | N |
| Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20 oC. |
| G | Little or no damage after 30 days constant exposure at 20oC. |
| F | Some effect after 7 days constant exposure at 20oC. |
| N | Not recommended. Immediate damage may occur. |
| Wide-Mouth Round Lab Quality Bottle (2105-series) | Wide-Mouth Round Economy Bottle (2187-series) | Large-Wide Mouth Round Bottle (2121-series) |
| Traditional reusable lab bottle for long-term use and critical applications | Lower cost, lighter weight, thinner wall for light-duty applications | Larger size, rugged lab quality construction |
![]() | ![]() | ![]() |
| PPCO bottle PP Closure | PPCO bottle PP Closure | PPCO bottle white pp closure |
| Milky white translucent | Milky white translucent | Milky white translucent |
| 30mL–1L | 30mL–1L | 2L–4L |
| Leakproof* | Leakproof* | Not liquid leakproof |
| No graduations | No graduations | No graduations |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40oC to 121oC |
| Autoclavable | Autoclavable | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free |
| Also available in bulk, unassembled | Also available in bulk, unassembled |
| Wide-Mouth Round Lab Quality Bottle (2105-series) | Wide-Mouth Round Economy Bottle (2187-series) | Large Wide-Mouth Round Bottle (2121-series) | |
| Acids, dilute or weak | E | E | E |
| Acids^ strong, concentrated | G | G | G |
| Alcohols, aliphatic | E | E | E |
| Aldehydes | G | G | G |
| Bases/alkali | E | E | E |
| Esters | G | G | G |
| Hydrocarbons, aliphatic | G | G | G |
| Hydrocarbons, aromatic | N | N | N |
| Hydrocarbons, halogenated | N | N | N |
| Ketones, aromatic | N | N | N |
| Oxidizing agents, strong | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20 oC |
| G | Little or no damage after 30 days constant exposure at 20oC |
| F | Some effect after 7 days constant exposure at 20oC |
| N | Not recommended. Immediate damage may occur. |
| Narrow-Mouth Square Lab Quality Bottle (2016-series) | Wide-Mouth Square Lab Quality Bottle (2110-series) | Large Wide-Mouth Square Bottle (2122-series) |
| Space-saving square shape, narrow mouth, graduated | Space-saving square shape, wide mouth | Space-saving square shape, wide mouth, large capacity, molded hand grip |
![]() | ![]() | ![]() |
| PPCO bottle/ PP Closure | PPCO bottle/ PP Closure | PPCO bottle/ white PP closure |
| Milky white translucent | Milky white translucent | Milky white translucent |
| 30mL–1L | 60mL–1L | 4L |
| L* | Leakproof* | Not liquid leakproof |
| Molded graduations | No graduations | No graduations |
| Narrow mouth | Wide mouth | Wide mouth |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -40°C to 121°C |
| Autoclavable | Autoclavable | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free |
| Narrow-Mouth Square Lab Quality Bottle (2016-series) | Wide-Mouth Square Lab Quality Bottle (2187-series) | Large Wide-Mouth Square Bottle (2122-series) | |
| Acids, dilute or weak | E | E | E |
| Acids^ strong, concentrated | G | G | G |
| Alcohols, aliphatic | E | E | E |
| Aldehydes | G | G | G |
| Bases/alkali | E | E | E |
| Esters | G | G | G |
| Hydrocarbons, aliphatic | G | G | G |
| Hydrocarbons, aromatic | N | N | N |
| Hydrocarbons, halogenated | N | N | N |
| Ketones, aromatic | N | N | N |
| Oxidizing agents, strong | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20oC. |
| G | Little or no damage after 30 days constant exposure at 20oC. |
| F | Some effect after 7 days constant exposure at 20oC. |
| N | Not recommended. Immediate damage may occur. |
For assistance choosing products appropriate for your application, please speak with a Nalgene Technical Support Representative team by phone at +1.585.586.8800 or (1.800.625.4327 US toll free), or email your request to
technicalsupport@thermofisher.com.
In Austria, France, Germany, Ireland, Switzerland and the United Kingdom please contact technical support by phone at +800 1234 9696 (toll free) or +49.6184.90.6321, or email your request to techsupport.labproducts.eu@thermofisher.com.
Regulatory Support: For regulatory documentation of product or material claims, please contact Nalgene Regulatory Support at RocRegSupport@thermofisher.com
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.